Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023991.1 | NP_076481.1 | 2 | 168 | ||
GenBank mRNA | U12149.1 | - | 2 | 353 | ||
Z29486.1 | - | 2 | 168 |