Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPP1 SIRNA Assays ›
          Gene Symbol:
          SPP1
          Gene Name:
          secreted phosphoprotein 1
          Gene Aliases:
          BNSP, BSPI, ETA-1, OPN
          Chromosome Location:
          Chr.4: 87975650 - 87983411 on Build GRCh38
          UniGene ID:
          Hs.313
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13377

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SPP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000582.2 NP_000573.1 5 354
          NM_001040058.1 NP_001035147.1 6 396
          NM_001040060.1 NP_001035149.1 5 315
          NM_001251829.1 NP_001238758.1 4 273
          NM_001251830.1 NP_001238759.1 7 585
          XM_017008564.1 XP_016864053.1 4 822
          GenBank mRNA AB209987.1 - 3 108
          AB469789.1 - 3 108
          AF052124.1 - 5 276
          AK057738.1 - 6 332
          AK075463.1 - 6 332
          AK290090.1 - 5 251
          AK290104.1 - 6 333
          AK295491.1 - 7 520
          AK296035.1 - 5 251
          AK315461.1 - 5 284
          BC007016.1 - 5 290 4052438
          BC017387.1 - 6 336 3828885
          BC022844.1 - 5 291 4284921
          BC093033.1 - 5 307 5288027
          BF699765.1 - 5 355
          BX648003.1 - 5 269
          D28759.1 - 5 231
          D28760.1 - 4 189
          D28761.1 - 4 150
          DA419665.1 - 5 354
          DQ839491.1 - 5 231
          DQ846870.1 - 5 231
          DQ846871.1 - 5 231
          HQ874460.1 - 2 69
          HQ874461.1 - 3 84
          J04765.1 - 5 297
          JF412666.1 - 5 233
          JF412667.1 - 5 313
          KR062183.1 - 7 520
          X13694.1 - 6 298

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          inflammatory response
          cell adhesion
          embryo implantation
          extracellular matrix disassembly
          extracellular matrix organization
          biomineral tissue development
          response to vitamin D
          positive regulation of bone resorption
          decidualization
          response to steroid hormone
          negative regulation of collateral sprouting of intact axon in response to injury
          cytokine activity
          protein binding
          extracellular matrix binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6b8df48494-rmf8w:80/100.66.134.75:80.
          git-commit: 2c12d7358bbebd854ca1589f3ab2f515852a3cea
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0