Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s132298
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
265
6921326
FQ211494.1
-
265
FQ211970.1
-
261
FQ212579.1
-
261
FQ212640.1
-
261
FQ212691.1
-
261
FQ212789.1
-
265
FQ212869.1
-
265
FQ213028.1
-
261
FQ213029.1
-
259
FQ213091.1
-
261
FQ213115.1
-
267
FQ213200.1
-
264
FQ213216.1
-
261
FQ213310.1
-
154
FQ213366.1
-
260
FQ213496.1
-
264
FQ213520.1
-
265
FQ213526.1
-
261
FQ213581.1
-
261
FQ213598.1
-
261
FQ213637.1
-
261
FQ213695.1
-
261
FQ213703.1
-
261
FQ213760.1
-
261
FQ213762.1
-
266
FQ213913.1
-
261
FQ213941.1
-
265
FQ213981.1
-
262
FQ213985.1
-
155
FQ214059.1
-
265
FQ214181.1
-
265
FQ214262.1
-
265
FQ214491.1
-
261
FQ214500.1
-
261
FQ214519.1
-
261
FQ214525.1
-
261
FQ214540.1
-
265
FQ215159.1
-
265
FQ215317.1
-
261
FQ215600.1
-
265
FQ216397.1
-
261
FQ216471.1
-
262
FQ218091.1
-
261
FQ220250.1
-
261
FQ220325.1
-
261
FQ220698.1
-
265
FQ220779.1
-
261
FQ222184.1
-
264
FQ223357.1
-
265
FQ225272.1
-
261
FQ226994.1
-
265
FQ229228.1
-
261
FQ229580.1
-
265
FQ229790.1
-
261
FQ229905.1
-
260
FQ231022.1
-
265
FQ231702.1
-
265
FQ233337.1
-
264
FQ234377.1
-
265
FQ235266.1
-
262
M17069.1
-
263
M19312.1
-
244
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0