Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017313.3 | NP_059009.2 | 2 | 504 | ||
XM_006241357.2 | XP_006241419.1 | 3 | 673 | |||
XM_006241358.2 | XP_006241420.1 | 3 | 678 | |||
XM_006241359.2 | XP_006241421.1 | 3 | 1682 | |||
XM_006241360.2 | XP_006241422.1 | 3 | 668 | |||
XM_008765382.1 | XP_008763604.1 | 2 | 730 | |||
XM_008765383.1 | XP_008763605.1 | 1 | 43 | |||
GenBank mRNA | BC127500.1 | - | 2 | 507 | 7457420 | |
U19181.1 | - | 2 | 497 |