Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTA1 SIRNA Assays ›
Gene Symbol:
Acta1
Gene Name:
actin, alpha 1, skeletal muscle
Gene Aliases:
-
Chromosome Location:
Chr.19: 56674072 - 56677084 on Build Rnor_6.0
UniGene ID:
Rn.82732
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s131417
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acta1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019212.2
NP_062085.1
3
487
GenBank mRNA
BC061974.1
-
3
487
5623909
FQ214583.1
-
3
496
FQ214585.1
-
3
498
FQ214598.1
-
3
496
FQ214600.1
-
3
496
FQ214603.1
-
3
496
FQ214610.1
-
1
41
FQ214623.1
-
3
496
FQ214640.1
-
3
496
FQ214659.1
-
3
498
FQ214676.1
-
3
496
FQ214680.1
-
3
496
FQ214697.1
-
3
496
FQ214734.1
-
3
496
FQ214743.1
-
3
496
FQ214756.1
-
3
496
FQ214761.1
-
3
495
FQ214779.1
-
3
496
FQ214785.1
-
3
494
FQ214786.1
-
3
496
FQ214807.1
-
3
496
FQ214814.1
-
3
496
FQ214822.1
-
3
496
FQ214831.1
-
3
496
FQ214832.1
-
3
496
FQ214839.1
-
3
499
FQ214841.1
-
3
496
FQ214842.1
-
3
496
FQ214857.1
-
3
500
FQ214858.1
-
3
496
FQ214859.1
-
3
496
FQ214860.1
-
3
496
FQ214862.1
-
3
496
FQ214866.1
-
3
496
FQ214883.1
-
3
495
FQ214904.1
-
3
496
FQ214905.1
-
3
495
FQ214916.1
-
3
496
FQ214921.1
-
3
496
FQ214936.1
-
3
496
FQ214948.1
-
3
496
FQ214949.1
-
3
496
FQ214957.1
-
3
496
FQ214969.1
-
3
496
FQ214975.1
-
3
498
FQ214976.1
-
3
496
FQ214978.1
-
3
496
FQ214987.1
-
3
496
FQ214990.1
-
3
496
FQ214993.1
-
3
496
FQ215012.1
-
3
496
FQ215025.1
-
3
496
FQ215038.1
-
3
496
FQ215044.1
-
3
496
FQ215058.1
-
3
496
FQ215089.1
-
3
496
FQ215112.1
-
3
496
FQ215136.1
-
3
496
FQ215149.1
-
3
496
FQ215153.1
-
3
496
FQ215158.1
-
3
494
FQ215173.1
-
3
496
FQ215185.1
-
3
495
FQ215204.1
-
3
494
FQ215212.1
-
3
496
FQ215225.1
-
3
496
FQ215241.1
-
3
496
FQ215249.1
-
3
497
FQ215253.1
-
3
496
FQ215256.1
-
3
496
FQ215275.1
-
3
496
FQ215277.1
-
3
497
FQ215280.1
-
3
496
FQ215282.1
-
3
496
FQ215298.1
-
3
498
FQ215324.1
-
3
496
FQ215333.1
-
3
496
FQ215342.1
-
3
496
FQ215346.1
-
3
496
FQ215366.1
-
3
496
FQ215380.1
-
3
496
FQ215381.1
-
3
496
FQ215393.1
-
3
496
FQ215396.1
-
3
496
FQ215398.1
-
3
496
FQ215402.1
-
3
496
FQ215403.1
-
3
496
FQ215405.1
-
3
496
FQ215414.1
-
3
498
FQ215424.1
-
3
496
FQ215431.1
-
3
496
FQ215433.1
-
3
495
FQ215434.1
-
3
498
FQ215438.1
-
3
496
FQ215446.1
-
3
496
FQ215449.1
-
3
498
FQ215454.1
-
3
496
FQ215464.1
-
3
496
FQ215465.1
-
3
496
FQ215468.1
-
3
496
FQ215470.1
-
3
496
FQ215474.1
-
3
496
FQ215483.1
-
3
496
FQ215488.1
-
3
496
FQ215518.1
-
3
505
FQ215523.1
-
3
496
FQ215537.1
-
3
498
FQ215554.1
-
3
496
FQ215567.1
-
3
496
FQ215572.1
-
3
496
FQ215593.1
-
3
496
FQ215618.1
-
3
496
FQ215628.1
-
3
496
FQ215631.1
-
1
110
FQ215655.1
-
3
494
FQ215659.1
-
3
495
FQ215673.1
-
3
496
FQ215674.1
-
3
496
FQ215690.1
-
3
496
FQ215692.1
-
3
496
FQ215785.1
-
3
496
FQ215790.1
-
3
498
FQ215799.1
-
3
496
FQ215809.1
-
3
496
FQ215813.1
-
3
496
FQ215835.1
-
3
496
FQ215836.1
-
3
496
FQ215839.1
-
3
496
FQ215840.1
-
3
496
FQ215841.1
-
3
496
FQ215851.1
-
3
496
FQ215864.1
-
3
497
FQ215866.1
-
3
498
FQ215888.1
-
3
495
FQ215893.1
-
3
497
FQ215896.1
-
3
496
FQ215907.1
-
3
498
FQ215950.1
-
3
496
FQ215964.1
-
3
496
FQ215972.1
-
3
496
FQ216016.1
-
3
496
FQ216035.1
-
3
496
FQ216043.1
-
3
496
FQ216050.1
-
3
496
FQ216066.1
-
3
494
FQ216138.1
-
3
496
FQ216139.1
-
3
496
FQ216168.1
-
3
494
FQ216196.1
-
3
496
FQ216224.1
-
3
496
FQ216246.1
-
3
496
FQ216276.1
-
3
496
FQ216278.1
-
3
498
FQ216287.1
-
3
494
FQ216291.1
-
3
496
FQ216293.1
-
3
496
FQ216305.1
-
3
496
FQ216333.1
-
3
496
FQ216350.1
-
3
496
FQ216352.1
-
3
496
FQ216360.1
-
3
496
FQ216369.1
-
3
498
FQ216403.1
-
3
496
FQ216430.1
-
3
496
FQ216509.1
-
3
494
FQ216586.1
-
3
496
FQ216603.1
-
3
496
FQ216606.1
-
3
496
FQ216607.1
-
3
492
FQ216610.1
-
3
494
FQ216620.1
-
3
496
FQ216635.1
-
3
494
FQ216639.1
-
3
496
FQ216644.1
-
3
496
FQ216664.1
-
3
494
FQ216675.1
-
3
496
FQ216676.1
-
3
496
FQ216692.1
-
3
496
FQ216703.1
-
3
496
FQ216713.1
-
3
496
FQ216721.1
-
3
496
FQ216723.1
-
3
496
FQ216725.1
-
3
492
FQ216739.1
-
3
496
FQ216762.1
-
3
494
FQ216767.1
-
3
496
FQ216776.1
-
3
496
FQ216777.1
-
3
496
FQ216779.1
-
3
498
FQ216793.1
-
3
496
FQ216799.1
-
3
494
FQ216801.1
-
3
496
FQ216814.1
-
3
490
FQ216819.1
-
3
494
FQ216916.1
-
3
496
FQ216918.1
-
1
139
FQ216961.1
-
3
496
FQ216994.1
-
3
496
FQ217061.1
-
3
498
FQ217073.1
-
3
499
FQ217091.1
-
3
495
FQ217114.1
-
3
496
FQ217122.1
-
3
496
FQ217167.1
-
3
496
FQ217173.1
-
3
496
FQ217188.1
-
3
496
FQ217194.1
-
3
496
FQ217273.1
-
3
495
FQ217280.1
-
3
616
FQ217327.1
-
3
496
FQ217329.1
-
3
496
FQ217347.1
-
3
496
FQ217353.1
-
3
496
FQ217367.1
-
3
496
FQ217397.1
-
3
496
FQ217441.1
-
3
496
FQ217443.1
-
3
498
FQ217475.1
-
3
496
FQ217525.1
-
3
496
FQ217535.1
-
3
496
FQ217572.1
-
3
496
FQ217593.1
-
3
496
FQ217602.1
-
3
496
FQ217615.1
-
3
496
FQ217629.1
-
3
496
FQ217641.1
-
3
493
FQ217762.1
-
3
496
FQ217826.1
-
3
496
FQ217884.1
-
3
494
FQ217893.1
-
3
496
FQ217905.1
-
3
496
FQ217908.1
-
3
496
FQ217937.1
-
3
496
FQ217969.1
-
3
498
FQ217990.1
-
3
496
FQ218019.1
-
3
496
FQ218034.1
-
3
496
FQ223563.1
-
3
496
FQ223681.1
-
3
498
FQ223686.1
-
3
496
FQ223702.1
-
3
496
FQ223727.1
-
1
194
FQ223898.1
-
3
496
FQ223914.1
-
3
496
FQ223926.1
-
3
496
FQ223929.1
-
3
496
FQ224009.1
-
3
496
FQ224082.1
-
3
496
FQ224141.1
-
3
497
FQ224167.1
-
3
496
FQ224359.1
-
3
496
FQ224377.1
-
3
496
FQ224419.1
-
3
496
FQ224552.1
-
3
496
FQ224691.1
-
3
494
FQ224777.1
-
3
496
FQ224840.1
-
3
494
FQ224842.1
-
3
496
FQ224862.1
-
3
496
FQ224869.1
-
3
496
FQ224886.1
-
3
496
FQ224890.1
-
3
494
FQ224902.1
-
3
496
FQ224929.1
-
3
494
FQ224936.1
-
3
496
FQ224942.1
-
3
498
FQ224946.1
-
1
24
FQ224948.1
-
3
498
FQ224962.1
-
3
496
FQ224971.1
-
3
496
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to mechanical stimulus
response to extracellular stimulus
response to lithium ion
cell growth
skeletal muscle thin filament assembly
skeletal muscle fiber adaptation
response to steroid hormone
skeletal muscle fiber development
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0