Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277450.1 | NP_001264379.1 | 3 | 483 | ||
NM_019191.2 | NP_062064.1 | 3 | 511 | |||
XM_006254946.2 | XP_006255008.1 | 3 | 224 | |||
GenBank mRNA | AB010147.1 | - | 2 | 285 | ||
AB017912.1 | - | 3 | 377 | |||
AF056001.1 | - | 3 | 411 | |||
BC127497.1 | - | 3 | 355 | 8364718 | ||
FQ230791.1 | - | 3 | 349 |