Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPSA SIRNA Assays ›
          Gene Symbol:
          Rpsa
          Gene Name:
          ribosomal protein SA
          Gene Aliases:
          Lamr1
          Chromosome Location:
          Chr.8: 128806056 - 128809987 on Build Rnor_6.0
          UniGene ID:
          Rn.121972
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s130986

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpsa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017138.2 NP_058834.1 4 475
          XM_008766672.1 XP_008764894.1 4 548
          GenBank mRNA BC060578.1 - 4 493 6919235
          D25224.1 - 4 476
          FJ386454.1 - 3 456
          FQ209463.1 - 4 489
          FQ209469.1 - 4 489
          FQ209490.1 - 4 490
          FQ209667.1 - 4 489
          FQ209729.1 - 4 489
          FQ211189.1 - 4 489
          FQ212660.1 - 4 489
          FQ212893.1 - 4 489
          FQ213275.1 - 4 489
          FQ213317.1 - 4 489
          FQ213334.1 - 4 489
          FQ213623.1 - 4 492
          FQ213835.1 - 4 490
          FQ213894.1 - 4 489
          FQ215242.1 - 4 489
          FQ215292.1 - 4 490
          FQ216063.1 - 4 489
          FQ216085.1 - 4 489
          FQ216130.1 - 4 489
          FQ216245.1 - 4 489
          FQ216788.1 - 4 491
          FQ216997.1 - 4 489
          FQ217664.1 - 4 489
          FQ219586.1 - 4 489
          FQ219785.1 - 4 492
          FQ220012.1 - 4 489
          FQ220017.1 - 4 487
          FQ220075.1 - 4 490
          FQ220084.1 - 4 492
          FQ220126.1 - 4 489
          FQ220134.1 - 4 489
          FQ220171.1 - 4 488
          FQ220184.1 - 4 489
          FQ220347.1 - 4 491
          FQ220461.1 - 4 470
          FQ220468.1 - 4 489
          FQ220661.1 - 4 492
          FQ220822.1 - 4 489
          FQ220904.1 - 4 490
          FQ220964.1 - 4 489
          FQ221103.1 - 4 489
          FQ221334.1 - 4 509
          FQ221443.1 - 4 490
          FQ221672.1 - 4 489
          FQ222093.1 - 4 491
          FQ222280.1 - 4 489
          FQ222542.1 - 4 491
          FQ222757.1 - 4 487
          FQ223223.1 - 4 489
          FQ224747.1 - 4 489
          FQ225510.1 - 4 490
          FQ226054.1 - 4 489
          FQ226408.1 - 4 489
          FQ227056.1 - 4 489
          FQ227293.1 - 4 489
          FQ228002.1 - 4 489
          FQ228212.1 - 4 490
          FQ229290.1 - 4 491
          FQ229305.1 - 4 491
          FQ229367.1 - 4 490
          FQ229516.1 - 4 489
          FQ229579.1 - 4 489
          FQ229598.1 - 4 490
          FQ229712.1 - 4 489
          FQ229787.1 - 4 489
          FQ229850.1 - 4 491
          FQ229859.1 - 4 487
          FQ229890.1 - 4 489
          FQ229918.1 - 4 509
          FQ229970.1 - 4 489
          FQ230088.1 - 4 489
          FQ230180.1 - 4 490
          FQ230258.1 - 4 488
          FQ231622.1 - 4 487
          FQ232305.1 - 4 491
          FQ232568.1 - 4 490
          FQ233068.1 - 4 489
          FQ233112.1 - 4 489
          FQ233340.1 - 4 491
          FQ233547.1 - 4 490
          FQ233919.1 - 4 489
          FQ234511.1 - 4 489
          FQ234742.1 - 4 489
          FQ234772.1 - 4 489

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          rRNA export from nucleus
          translation
          single organismal cell-cell adhesion
          epithelial cell differentiation
          structural constituent of ribosome
          laminin receptor activity
          structural molecule activity
          ribosome binding
          laminin binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0