Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017118.1 | NP_058814.1 | 607 | |||
GenBank mRNA | BC098068.1 | - | 607 | 7462537 | ||
FQ229527.1 | - | 290 | ||||
U10861.1 | - | 5 | 296 | |||
U53859.1 | - | 543 |
Gene Symbol: LOC100911363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748836.3 | XP_003748884.1 | 8 | 656 | ||
XM_008759263.1 | XP_008757485.1 | 8 | 858 |
Gene Symbol: LOC100912380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228796.2 | XP_006228858.2 | 7 | 383 |