Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEDD4 SIRNA Assays ›
          Gene Symbol:
          Nedd4
          Gene Name:
          neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase
          Gene Aliases:
          Nedd4a
          Chromosome Location:
          Chr.8: 79323408 - 79408842 on Build Rnor_6.0
          UniGene ID:
          Rn.99540
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s129933

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nedd4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012986.1 NP_037118.1 6 502
          GenBank mRNA CK365888.1 - 6 502
          U50842.1 - 6 428

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          adaptive immune response
          outflow tract morphogenesis
          endocardial cushion development
          protein monoubiquitination
          protein targeting to lysosome
          immune response
          lysosomal transport
          neuromuscular junction development
          negative regulation of sodium ion transport
          negative regulation of transcription from RNA polymerase II promoter in response to UV-induced DNA damage
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to denervation involved in regulation of muscle adaptation
          protein ubiquitination
          transmission of virus
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          neuron projection development
          receptor internalization
          receptor catabolic process
          cellular response to UV
          regulation of ion transmembrane transport
          T cell activation
          regulation of membrane potential
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          glucocorticoid receptor signaling pathway
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          development involved in symbiotic interaction
          positive regulation of protein catabolic process
          positive regulation of nucleocytoplasmic transport
          blood vessel morphogenesis
          regulation of dendrite morphogenesis
          regulation of synapse organization
          progesterone receptor signaling pathway
          protein K63-linked ubiquitination
          regulation of potassium ion transmembrane transporter activity
          ubiquitin-protein transferase activity
          protein binding
          protein C-terminus binding
          ligase activity
          sodium channel inhibitor activity
          protein domain specific binding
          beta-2 adrenergic receptor binding
          ubiquitin binding
          phosphoserine binding
          phosphothreonine binding
          ubiquitin protein ligase activity
          RNA polymerase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0