Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GDNF SIRNA Assays ›
          Gene Symbol:
          Gdnf
          Gene Name:
          glial cell derived neurotrophic factor
          Gene Aliases:
          gndf
          Chromosome Location:
          Chr.2: 56887546 - 56912964 on Build Rnor_6.0
          UniGene ID:
          Rn.53970
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s129835

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gdnf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019139.1 NP_062012.1 2, 3 209
          XM_006232010.2 XP_006232072.1 2, 3 650
          XM_008760756.1 XP_008758978.1 1, 2 1487
          GenBank mRNA AF205713.1 - 2, 3 213
          AF497634.1 - 1, 2 137
          EU068469.1 - 1, 2 175
          EU068470.1 - 1, 2 160
          EU068471.1 - 1, 2 160
          EU068472.1 - 1, 2 160
          L15305.1 - 2, 3 209
          S75583.1 - 1, 2 160

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          neural crest cell migration
          organ induction
          postsynaptic membrane organization
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          nervous system development
          peripheral nervous system development
          cell proliferation
          positive regulation of cell proliferation
          male gonad development
          response to wounding
          regulation of gene expression
          postganglionic parasympathetic fiber development
          neuron differentiation
          peristalsis
          midbrain development
          neuron projection development
          positive regulation of monooxygenase activity
          negative regulation of neuron apoptotic process
          positive regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          mRNA stabilization
          enteric nervous system development
          sympathetic nervous system development
          regulation of dopamine uptake involved in synaptic transmission
          retina development in camera-type eye
          ureteric bud formation
          regulation of morphogenesis of a branching structure
          cellular response to dexamethasone stimulus
          regulation of ureteric bud formation
          positive regulation of ureteric bud formation
          positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          positive regulation of branching involved in ureteric bud morphogenesis
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          growth factor activity
          glial cell-derived neurotrophic factor receptor binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0