Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031549.2 | NP_113737.1 | 2, 3 | 236 | ||
GenBank mRNA | BC061770.1 | - | 2, 3 | 236 | 5621257 | |
FQ212757.1 | - | 2, 3 | 261 | |||
FQ219883.1 | - | 2, 3 | 259 | |||
FQ220131.1 | - | 2, 3 | 260 | |||
FQ220322.1 | - | 2, 3 | 258 | |||
FQ220861.1 | - | 2, 3 | 261 | |||
FQ220863.1 | - | 2, 3 | 261 | |||
FQ223284.1 | - | 2, 3 | 260 | |||
FQ224487.1 | - | 2, 3 | 261 | |||
FQ229263.1 | - | 2, 3 | 259 | |||
FQ229406.1 | - | 2, 3 | 258 | |||
FQ229775.1 | - | 2, 3 | 260 | |||
FQ229807.1 | - | 2, 3 | 261 | |||
FQ234912.1 | - | 2, 3 | 256 | |||
M83107.1 | - | 2, 3 | 346 | |||
X64422.1 | - | 1, 2 | 184 | |||
X71070.1 | - | 2, 3 | 233 |