Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012686.2 | NP_036818.2 | 4 | 707 | ||
GenBank mRNA | BC092197.1 | - | 4 | 707 | 7313365 | |
D10666.1 | - | 4 | 715 |