Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191649.1 | NP_001178578.1 | 2 | 532 | ||
XM_006247175.2 | XP_006247237.1 | 2 | 541 |