Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012530.2 | NP_036662.1 | 3 | 424 | ||
GenBank mRNA | BC062058.1 | - | 3 | 375 | 5598226 | |
DN931989.1 | - | 3 | 423 | |||
FQ214594.1 | - | 3 | 400 | |||
FQ214655.1 | - | 3 | 400 | |||
FQ214750.1 | - | 3 | 400 | |||
FQ214825.1 | - | 2 | 221 | |||
FQ214931.1 | - | 3 | 401 | |||
FQ215369.1 | - | 2 | 298 | |||
FQ215410.1 | - | 3 | 400 | |||
FQ215629.1 | - | 1 | 17 | |||
FQ215927.1 | - | 2 | 253 | |||
FQ216321.1 | - | 2 | 252 | |||
FQ216507.1 | - | 3 | 395 | |||
FQ216611.1 | - | 1 | 17 | |||
FQ217421.1 | - | 2 | 265 | |||
FQ224848.1 | - | 2 | 251 | |||
FQ224918.1 | - | 3 | 401 | |||
M10140.1 | - | 3 | 397 | |||
M14864.1 | - | 1 | 70 |