Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016966.3 | NP_058662.2 | 2 | 417 | ||
GenBank mRNA | AK076815.1 | - | 2 | 373 | ||
AK147946.1 | - | 2 | 365 | |||
AK169684.1 | - | 2 | 373 | |||
BC086668.1 | - | 2 | 325 | 30633243 | ||
BC110673.1 | - | 2 | 359 | 6311873 | ||
BY290664.1 | - | 2 | 417 | |||
L21027.1 | - | 1 | 136 |
Gene Symbol: Gm6756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076985.1 | - | 1 | 48 | ||
BB616006.1 | - | 1 | 48 |