Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 8 | 1131 | ||
GenBank mRNA | AK034358.1 | - | 8 | 1163 | ||
AK036245.1 | - | 8 | 1207 | |||
AK045969.1 | - | 8 | 1165 | |||
AK048738.1 | - | 8 | 1117 | |||
AK079644.1 | - | 7 | 1062 | |||
AK082474.1 | - | 7 | 1076 | |||
AK133610.1 | - | 8 | 1132 | |||
AK134808.1 | - | 8 | 1192 | |||
AK161902.1 | - | 8 | 1192 | |||
BC132290.1 | - | 8 | 1107 | 40130567 | ||
BC138012.1 | - | 8 | 1107 | 8861031 |