Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291137.1 | NP_001278066.1 | 6 | 1111 | ||
NM_001291138.1 | NP_001278067.1 | 6 | 1111 | |||
GenBank mRNA | AK046067.1 | - | 6 | 1086 | ||
BC138377.1 | - | 6 | 1013 | 8861397 | ||
BC145260.1 | - | 6 | 1013 |