Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
Trpm3
Gene Name:
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases:
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
Chromosome Location:
Chr.19: 22138181 - 22989897 on Build GRCm38
UniGene ID:
Mm.131943
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s105261
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Trpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035239.2
NP_001030316.1
7
1035
NM_001035240.2
NP_001030317.1
7
1035
NM_001035241.2
NP_001030318.1
7
1035
NM_001035242.1
NP_001030319.1
7
1035
NM_001035243.2
NP_001030320.1
7
1035
NM_001035244.1
NP_001030321.1
8
1158
NM_001035245.1
NP_001030322.1
7
1083
NM_001035246.1
NP_001030323.1
8
1158
NM_177341.4
NP_796315.2
7
1102
GenBank mRNA
AJ544532.1
-
7
1035
AJ544533.1
-
7
1035
AJ544534.1
-
7
1035
AJ544535.1
-
7
1035
AJ544536.1
-
7
1035
AK044454.1
-
7
1104
AK044533.1
-
8
956
AK046908.1
-
8
991
AK051867.1
-
1
58
AK051934.1
-
8
956
AK052064.1
-
8
964
AK053673.1
-
7
920
AK082141.1
-
7
917
AK084402.1
-
8
963
AK142504.1
-
8
991
AK161909.1
-
8
991
AK173218.1
-
7
1106
BC037449.1
-
7
931
BC141400.1
-
7
1078
9056013
BC141412.1
-
7
1078
9056025
HM064407.1
-
7
1127
HM064408.1
-
7
1127
HM064409.1
-
7
1127
HM064410.1
-
8
1202
HM064411.1
-
7
1127
JF706722.1
-
7
1098
JX644970.1
-
7
1127
JX644971.1
-
7
1127
JX644972.1
-
7
1127
JX644973.1
-
7
1127
JX644974.1
-
7
1127
JX644975.1
-
8
1202
JX644976.1
-
7
1127
JX644977.1
-
7
1127
JX644978.1
-
7
1127
JX644979.1
-
8
1202
JX644980.1
-
8
1202
JX644981.1
-
8
1202
JX644982.1
-
8
1202
JX644983.1
-
7
1127
JX644984.1
-
7
1127
JX644985.1
-
7
1127
JX644986.1
-
7
1127
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191562.
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
cation transmembrane transport
cation channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
git-commit: a47d3bef29014c2544d0032e3b149846d456d521
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0