Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLGN1 SIRNA Assays ›
          Gene Symbol:
          Nlgn1
          Gene Name:
          neuroligin 1
          Gene Aliases:
          6330415N05Rik, BB179718, NL1, Nlg1, mKIAA1070
          Chromosome Location:
          Chr.3: 25431841 - 26332056 on Build GRCm38
          UniGene ID:
          Mm.316080
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s101246

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nlgn1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163387.1 NP_001156859.1 5 1516
          NM_138666.3 NP_619607.2 7 2543
          GenBank mRNA AK122433.1 - 7 2544
          AK138209.1 - 2 377
          BC098461.1 - 5 1516 30099449

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of respiratory gaseous exchange by neurological system process
          protein targeting
          cell adhesion
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          neuron cell-cell adhesion
          nervous system development
          synapse assembly
          positive regulation of circadian sleep/wake cycle, wakefulness
          synaptic vesicle targeting
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          neuron projection development
          positive regulation of synaptic transmission, GABAergic
          protein localization to synapse
          establishment of protein localization
          regulation of neuron differentiation
          synaptic vesicle transport
          rhythmic process
          cytoskeletal matrix organization at active zone
          modulation of synaptic transmission
          synapse organization
          protein homooligomerization
          protein heterotetramerization
          positive regulation of filopodium assembly
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          inhibitory postsynaptic potential
          long-term synaptic potentiation
          positive regulation of dendritic spine development
          negative regulation of dendritic spine morphogenesis
          terminal button organization
          synaptic vesicle clustering
          postsynaptic membrane assembly
          presynaptic membrane assembly
          AMPA glutamate receptor clustering
          NMDA glutamate receptor clustering
          neurexin clustering involved in presynaptic membrane assembly
          postsynaptic density protein 95 clustering
          receptor localization to synapse
          positive regulation of ruffle assembly
          positive regulation of synaptic vesicle endocytosis
          positive regulation of protein localization to synapse
          positive regulation of intracellular signal transduction
          positive regulation of synaptic vesicle exocytosis
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          positive regulation of synaptic vesicle clustering
          receptor activity
          protein binding
          PDZ domain binding
          neurexin family protein binding
          protein dimerization activity
          cell adhesion molecule binding
          carboxylic ester hydrolase activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0