Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 696 | |||
NR_110066.1 | 3 | 378 | ||||
NR_110067.1 | 4 | 634 | ||||
NR_110068.1 | 2 | 334 | ||||
NR_110069.1 | 3 | 526 | ||||
NR_110070.1 | 5 | 485 | ||||
GenBank mRNA | BG478902.1 | - | 379 | |||
BM544796.1 | - | 5 | 506 | |||
HY003139.1 | - | 3 | 350 |