Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105372476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136528.1 | 4 | 645 | |||
NR_136529.1 | 3 | 578 | ||||
GenBank mRNA | BX098599.1 | - | 4 | 619 |