Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEAT1 SIRNA Assays ›
          Gene Symbol:
          NEAT1
          Gene Name:
          nuclear paraspeckle assembly transcript 1 (non-protein coding)
          Gene Aliases:
          LINC00084, NCRNA00084, TncRNA, VINC
          Chromosome Location:
          Chr.11: 65422798 - 65445540 on Build GRCh38
          UniGene ID:
          Hs.523789
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n544966

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEAT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028272.1 1 2926
          NR_131012.1 1 2926
          GenBank mRNA AF001893.1 - 1 678
          AK027191.1 - 1 910
          EF177379.1 - 1 2926
          GQ859162.1 - 1 2926
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX457383.2 - 1 340 Chr. 11 - 65425383 - 65426255 [+] on Build GRCh38
          GenBankmRNA BG621344.1 - 1 135 Chr. 11 - 65425589 - 65426359 [+] on Build GRCh38
          GenBankmRNA CB963225.1 - 1 203 Chr. 11 - 65425523 - 65426287 [+] on Build GRCh38
          GenBankmRNA BF679881.1 - 1 20 Chr. 11 - 65425708 - 65426288 [+] on Build GRCh38
          GenBankmRNA BQ721690.1 - 1 124 Chr. 11 - 65425600 - 65426525 [+] on Build GRCh38
          GenBankmRNA BG674955.1 - 1 103 Chr. 11 - 65425622 - 65426434 [+] on Build GRCh38
          GenBankmRNA BY996298.1 - 1 107 Chr. 11 - 65425625 - 65426277 [+] on Build GRCh38
          GenBankmRNA BG696830.1 - 1 254 Chr. 11 - 65425470 - 65426261 [+] on Build GRCh38
          GenBankmRNA HX786077.1 - 1 135 Chr. 11 - 65425589 - 65426234 [+] on Build GRCh38
          GenBankmRNA CB854402.1 - 1 149 Chr. 11 - 65425586 - 65426184 [+] on Build GRCh38
          GenBankmRNA EL955070.1 - 1 105 Chr. 11 - 65425621 - 65426180 [+] on Build GRCh38
          GenBankmRNA CB155865.1 - 1 58 Chr. 11 - 65425666 - 65426262 [+] on Build GRCh38
          GenBankmRNA CD722075.1 - 1 109 Chr. 11 - 65425615 - 65426202 [+] on Build GRCh38
          GenBankmRNA BG113348.1 - 1 25 Chr. 11 - 65425699 - 65426357 [+] on Build GRCh38
          GenBankmRNA BP417015.1 - 1 45 Chr. 11 - 65425679 - 65426282 [+] on Build GRCh38
          GenBankmRNA DR423495.1 - 1 204 Chr. 11 - 65425524 - 65426245 [+] on Build GRCh38
          GenBankmRNA BG674297.1 - 1 200 Chr. 11 - 65425524 - 65426351 [+] on Build GRCh38
          GenBankmRNA AL547749.3 - 1 210 Chr. 11 - 65425515 - 65426391 [+] on Build GRCh38
          GenBankmRNA BI832781.1 - 1 49 Chr. 11 - 65425674 - 65426408 [+] on Build GRCh38
          GenBankmRNA HX787970.1 - 1 135 Chr. 11 - 65425589 - 65426234 [+] on Build GRCh38
          GenBankmRNA BG740944.1 - 1 206 Chr. 11 - 65425517 - 65426312 [+] on Build GRCh38
          GenBankmRNA CU445808.1 - 1 81 Chr. 11 - 65425654 - 65426228 [+] on Build GRCh38
          GenBankmRNA BU174238.1 - 1 136 Chr. 11 - 65425588 - 65426391 [+] on Build GRCh38
          GenBankmRNA BG573479.1 - 1 50 Chr. 11 - 65425674 - 65426375 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0