Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC01089 SIRNA Assays ›
          Gene Symbol:
          LINC01089
          Gene Name:
          long intergenic non-protein coding RNA 1089
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 121795267 - 121803484 on Build GRCh38
          UniGene ID:
          Hs.524804
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n544657

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC01089
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002809.2 5, 6 1018
          GenBank mRNA AK094482.1 - 1 646
          AK095700.1 - 1 1596
          BC038786.1 - 5, 6 1018
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD673695.1 - 2, 3 263 Chr. 12 - 121795486 - 121800559 [-] on Build GRCh38
          GenBankmRNA BF125696.1 - 4, 5 393 Chr. 12 - 121795926 - 121802901 [-] on Build GRCh38
          GenBankmRNA BI818784.1 - 4, 5 430 Chr. 12 - 121795804 - 121802933 [-] on Build GRCh38
          GenBankmRNA BG287319.1 - 178 Chr. - - [-] on Build
          GenBankmRNA BQ068043.1 - 1 691 Chr. 12 - 121796058 - 121796924 [-] on Build GRCh38
          GenBankmRNA BG035222.1 - 482 Chr. - - [-] on Build
          GenBankmRNA BM264197.1 - 1 418 Chr. 12 - 121796209 - 121796651 [-] on Build GRCh38
          GenBankmRNA BE266310.1 - 3, 4 489 Chr. 12 - 121796116 - 121802905 [-] on Build GRCh38
          GenBankmRNA BG393185.1 - 4, 5 429 Chr. 12 - 121796145 - 121802933 [-] on Build GRCh38
          GenBankmRNA BE796247.1 - 2, 3 198 Chr. 12 - 121795735 - 121801785 [-] on Build GRCh38
          GenBankmRNA AI634559.1 - 2, 3 277 Chr. 12 - 121795574 - 121801005 [-] on Build GRCh38
          GenBankmRNA HY000339.1 - 2, 3 269 Chr. 12 - 121796057 - 121800563 [-] on Build GRCh38
          GenBankmRNA BM470301.1 - 423 Chr. - - [-] on Build
          GenBankmRNA CN310936.1 - 3, 4 513 Chr. 12 - 121796221 - 121802920 [-] on Build GRCh38
          GenBankmRNA BX431441.2 - 430 Chr. - - [-] on Build
          GenBankmRNA BX424339.2 - 1 737 Chr. 12 - 121796032 - 121796969 [-] on Build GRCh38
          GenBankmRNA CD516035.1 - 4, 5 417 Chr. 12 - 121795815 - 121802920 [-] on Build GRCh38
          GenBankmRNA BI912454.1 - 184 Chr. - - [-] on Build
          GenBankmRNA CN398873.1 - 4, 5 439 Chr. 12 - 121795590 - 121802930 [-] on Build GRCh38
          GenBankmRNA BM698927.1 - 3, 4 511 Chr. 12 - 121796211 - 121802920 [-] on Build GRCh38
          GenBankmRNA CN398874.1 - 1 509 Chr. 12 - 121796125 - 121796742 [-] on Build GRCh38
          GenBankmRNA DA365821.1 - 3, 4 534 Chr. 12 - 121796210 - 121802943 [-] on Build GRCh38
          GenBankmRNA HY077241.1 - 2, 3 398 Chr. 12 - 121796162 - 121801970 [-] on Build GRCh38
          GenBankmRNA DT219909.1 - 3, 4 536 Chr. 12 - 121796095 - 121802945 [-] on Build GRCh38
          GenBankmRNA BM724477.1 - 1, 2 134 Chr. 12 - 121795731 - 121799958 [-] on Build GRCh38
          GenBankmRNA BG699859.1 - 4, 5 444 Chr. 12 - 121795399 - 121802942 [-] on Build GRCh38
          GenBankmRNA BX344455.2 - 4, 5 430 Chr. 12 - 121795769 - 121802930 [-] on Build GRCh38
          GenBankmRNA BG696176.1 - 3, 4 240 Chr. 12 - 121795693 - 121801947 [-] on Build GRCh38
          GenBankmRNA DT218208.1 - 4, 5 442 Chr. 12 - 121796041 - 121802945 [-] on Build GRCh38
          GenBankmRNA BM548548.1 - 126 Chr. - - [-] on Build
          GenBankmRNA BM922850.1 - 1 655 Chr. 12 - 121795892 - 121796864 [-] on Build GRCh38
          GenBankmRNA BE797083.1 - 4, 5 403 Chr. 12 - 121795879 - 121802906 [-] on Build GRCh38
          GenBankmRNA BE791253.1 - 253 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f76987d59-9ndr4:80/100.66.134.75:80.
          git-commit: fa11bf362e7d47d57dd0debebfa11e830f891645
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0