Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 1600020E01RIK SIRNA Assays ›
          Gene Symbol:
          1600020E01Rik
          Gene Name:
          RIKEN cDNA 1600020E01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 86527330 - 86564449 on Build GRCm38
          UniGene ID:
          Mm.349174
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n521193

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 1600020E01Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037960.1 3 260
          NR_037961.1 3 287
          GenBank mRNA AA162444.1 - 3 258
          AI644794.1 - 3 256
          AK005503.1 - 3 288
          BC029726.1 - 1777
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD768781.1 - 3 275 Chr. 6 - 86527319 - 86564449 [+] on Build GRCm38
          GenBankmRNA EH111877.1 - 2 143 Chr. 6 - 86527904 - 86564449 [+] on Build GRCm38
          GenBankmRNA BQ747927.1 - 224 Chr. - - [-] on Build
          GenBankmRNA BI415363.1 - 3 631 Chr. 6 - 86526752 - 86564332 [+] on Build GRCm38
          GenBankmRNA BF178438.1 - 2 190 Chr. 6 - 86527857 - 86564424 [+] on Build GRCm38
          GenBankmRNA CJ310987.1 - 2 93 Chr. 6 - 86527954 - 86564448 [+] on Build GRCm38
          GenBankmRNA CD769707.1 - 3 275 Chr. 6 - 86527319 - 86564449 [+] on Build GRCm38
          GenBankmRNA AA198552.1 - 3 253 Chr. 6 - 86527337 - 86564430 [+] on Build GRCm38
          GenBankmRNA BY705705.1 - 3 284 Chr. 6 - 86527404 - 86564448 [+] on Build GRCm38
          GenBankmRNA AA792604.1 - 3 222 Chr. 6 - 86527473 - 86563505 [+] on Build GRCm38
          GenBankmRNA BE198520.1 - 3 192 Chr. 6 - 86526247 - 86563489 [+] on Build GRCm38
          GenBankmRNA DV067145.1 - 306 Chr. - - [-] on Build
          GenBankmRNA CJ301254.1 - 2 88 Chr. 6 - 86527959 - 86564448 [+] on Build GRCm38
          GenBankmRNA CJ302153.1 - 1 83 Chr. 6 - 86532192 - 86564448 [+] on Build GRCm38
          GenBankmRNA BE849333.1 - 3 192 Chr. 6 - 86526247 - 86551042 [+] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0