Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 2410006H16RIK SIRNA Assays ›
          Gene Symbol:
          2410006H16Rik
          Gene Name:
          RIKEN cDNA 2410006H16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 62602877 - 62604806 on Build GRCm38
          UniGene ID:
          Mm.270874
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n519374

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 2410006H16Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_030738.1 5 347
          GenBank mRNA AK010427.1 - 5 348
          AK157519.1 - 3 1291
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CF612138.1 - 5 318 Chr. 11 - 62602943 - 62604809 [+] on Build GRCm38
          GenBankmRNA CA469348.1 - 384 Chr. - - [-] on Build
          GenBankmRNA W07913.1 - 202 Chr. - - [-] on Build
          GenBankmRNA CK795379.1 - 5 337 Chr. 11 - 62602901 - 62604809 [+] on Build GRCm38
          GenBankmRNA AV478545.1 - 5 323 Chr. 11 - 62602901 - 62604804 [+] on Build GRCm38
          GenBankmRNA AA087133.1 - 5 319 Chr. 11 - 62602903 - 62604801 [+] on Build GRCm38
          GenBankmRNA AV490079.1 - 5 320 Chr. 11 - 62602903 - 62604796 [+] on Build GRCm38
          GenBankmRNA AA939619.1 - 5 274 Chr. 11 - 62602948 - 62604804 [+] on Build GRCm38
          GenBankmRNA AV483941.1 - 5 331 Chr. 11 - 62602892 - 62604784 [+] on Build GRCm38
          GenBankmRNA W30208.1 - 328 Chr. - - [-] on Build
          GenBankmRNA AV576844.1 - 5 322 Chr. 11 - 62602902 - 62604804 [+] on Build GRCm38
          GenBankmRNA BY471947.1 - 5 347 Chr. 11 - 62602877 - 62604801 [+] on Build GRCm38
          GenBankmRNA AV459516.1 - 5 335 Chr. 11 - 62602892 - 62604809 [+] on Build GRCm38
          GenBankmRNA AV492656.1 - 5 325 Chr. 11 - 62602900 - 62604809 [+] on Build GRCm38
          GenBankmRNA AA052212.1 - 5 313 Chr. 11 - 62602910 - 62604745 [+] on Build GRCm38
          GenBankmRNA W34684.1 - 321 Chr. - - [-] on Build
          GenBankmRNA BY405332.1 - 5 290 Chr. 11 - 62602936 - 62604806 [+] on Build GRCm38
          GenBankmRNA BE647248.1 - 5 218 Chr. 11 - 62603006 - 62604809 [+] on Build GRCm38
          GenBankmRNA AV500889.1 - 5 226 Chr. 11 - 62602998 - 62604778 [+] on Build GRCm38
          GenBankmRNA CJ176622.1 - 5 344 Chr. 11 - 62602880 - 62604778 [+] on Build GRCm38
          GenBankmRNA W29476.1 - 331 Chr. - - [-] on Build
          GenBankmRNA AV491046.1 - 5 336 Chr. 11 - 62602890 - 62604777 [+] on Build GRCm38
          GenBankmRNA AI060452.1 - 5 333 Chr. 11 - 62602891 - 62604782 [+] on Build GRCm38
          GenBankmRNA AV508300.1 - 5 331 Chr. 11 - 62602893 - 62604804 [+] on Build GRCm38
          GenBankmRNA BY709861.1 - 5 343 Chr. 11 - 62602884 - 62604806 [+] on Build GRCm38
          GenBankmRNA DV054738.1 - 368 Chr. - - [-] on Build
          GenBankmRNA AV457388.1 - 5 347 Chr. 11 - 62602883 - 62604778 [+] on Build GRCm38
          GenBankmRNA W13601.1 - 5 321 Chr. 11 - 62602904 - 62604767 [+] on Build GRCm38
          GenBankmRNA CF587018.1 - 4 206 Chr. 11 - 62603241 - 62604809 [+] on Build GRCm38
          GenBankmRNA BE138252.1 - 5 340 Chr. 11 - 62602895 - 62604782 [+] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0