Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933439C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015585.2 | 1, 2 | 1210 | |||
GenBank mRNA | AK011602.1 | - | 1, 2 | 743 | ||
AK017115.1 | - | 1, 2 | 1211 | |||
AK030920.1 | - | 1238 | ||||
AK051100.1 | - | 1, 2 | 228 | |||
AK163429.1 | - | 1, 2 | 1206 | |||
AK169756.1 | - | 1, 2 | 1240 | |||
BC099591.1 | - | 1, 2 | 457 |