Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100287592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104653.1 | 1 | 152 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX108736.1 | - | 1 | 110 | Chr. 5 - 50969411 - 50970145 [-] on Build GRCh38 |