Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100507291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121608.1 | 3 | 333 | |||
NR_121609.1 | 3 | 333 | ||||
GenBank mRNA | BX098847.1 | - | 3 | 327 | ||
DA364121.1 | - | 3 | 296 | |||
DB034152.1 | - | 3 | 333 | |||
DB446595.1 | - | 3 | 334 |