Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEAT1 SIRNA Assays ›
          Gene Symbol:
          NEAT1
          Gene Name:
          nuclear paraspeckle assembly transcript 1 (non-protein coding)
          Gene Aliases:
          LINC00084, NCRNA00084, TncRNA, VINC
          Chromosome Location:
          Chr.11: 65422798 - 65445540 on Build GRCh38
          UniGene ID:
          Hs.523789
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n509902

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEAT1
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028272.1
          1 3320
          NR_131012.1
          1 3320
          GenBank mRNA AF001893.1 -
          1 1072
          AF080092.1 - LIT3313
          TncRNA
          1 67
          AF508303.1 - LIT1827
          TncRNA
          1 67
          AK027191.1 -
          1 1304
          EF177379.1 -
          1 3320
          GQ859162.1 -
          1 3320
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA352268.1 - 1 192 Chr. 11 - 65425926 - 65426245 [+] on Build GRCh38
          GenBankmRNA AA634795.1 - 1 140 Chr. 11 - 65425978 - 65426355 [+] on Build GRCh38
          GenBankmRNA BE178454.1 - 1 105 Chr. 11 - 65426013 - 65426394 [+] on Build GRCh38
          GenBankmRNA BE178376.1 - 1 115 Chr. 11 - 65426007 - 65426385 [+] on Build GRCh38
          GenBankmRNA BM999234.1 - 1 177 Chr. 11 - 65425945 - 65426469 [+] on Build GRCh38
          GenBankmRNA AJ709584.1 - 1 100 Chr. 11 - 65426018 - 65426518 [+] on Build GRCh38
          GenBankmRNA AV696579.2 - 1 80 Chr. 11 - 65426038 - 65426471 [+] on Build GRCh38
          GenBankmRNA BE708149.1 - 1 48 Chr. 11 - 65426070 - 65426384 [+] on Build GRCh38
          GenBankmRNA AV684892.1 - 1 80 Chr. 11 - 65426038 - 65426474 [+] on Build GRCh38
          GenBankmRNA BE178398.1 - 1 110 Chr. 11 - 65426008 - 65426385 [+] on Build GRCh38
          GenBankmRNA BM796270.1 - 1 238 Chr. 11 - 65425880 - 65426480 [+] on Build GRCh38
          GenBankmRNA CB854402.1 - 1 543 Chr. 11 - 65425586 - 65426184 [+] on Build GRCh38
          GenBankmRNA AA483662.1 - 1 183 Chr. 11 - 65425942 - 65426223 [+] on Build GRCh38
          GenBankmRNA AJ710291.1 - 1 393 Chr. 11 - 65425737 - 65426279 [+] on Build GRCh38
          GenBankmRNA BF351392.1 - 169 Chr. - - [-] on Build
          GenBankmRNA AI028143.1 - 2 223 Chr. 11 - 65368058 - 65426222 [+] on Build GRCh38
          GenBankmRNA BM727919.1 - 1 127 Chr. 11 - 65425991 - 65426260 [+] on Build GRCh38
          GenBankmRNA CB984347.1 - 121 Chr. - - [-] on Build
          GenBankmRNA BE178389.1 - 1 123 Chr. 11 - 65426016 - 65426385 [+] on Build GRCh38
          GenBankmRNA BE166949.1 - 1 146 Chr. 11 - 65425973 - 65426304 [+] on Build GRCh38
          GenBankmRNA BE840456.1 - 1 87 Chr. 11 - 65426062 - 65426370 [+] on Build GRCh38
          GenBankmRNA BE840467.1 - 1 108 Chr. 11 - 65426019 - 65426231 [+] on Build GRCh38
          GenBankmRNA BE840544.1 - 1 108 Chr. 11 - 65426019 - 65426231 [+] on Build GRCh38
          GenBankmRNA CD240095.1 - 1 385 Chr. 11 - 65425784 - 65426507 [+] on Build GRCh38
          GenBankmRNA CD679171.1 - 1 35 Chr. 11 - 65426083 - 65426524 [+] on Build GRCh38
          GenBankmRNA CB162181.1 - 1 289 Chr. 11 - 65425829 - 65426354 [+] on Build GRCh38
          GenBankmRNA AA053611.1 - 1 54 Chr. 11 - 65426064 - 65426462 [+] on Build GRCh38
          GenBankmRNA AL547749.3 - 1 615 Chr. 11 - 65425515 - 65426391 [+] on Build GRCh38
          GenBankmRNA BE177865.1 - 426 Chr. - - [-] on Build
          GenBankmRNA BE842714.1 - 1 351 Chr. 11 - 65425773 - 65426304 [+] on Build GRCh38
          GenBankmRNA DR421841.1 - 1 47 Chr. 11 - 65426074 - 65426532 [+] on Build GRCh38
          GenBankmRNA AL080053.2 - 1 77 Chr. 11 - 65426041 - 65426402 [+] on Build GRCh38
          GenBankmRNA BG740944.1 - 1 600 Chr. 11 - 65425517 - 65426312 [+] on Build GRCh38
          GenBankmRNA AA045143.1 - 1 45 Chr. 11 - 65426081 - 65426526 [+] on Build GRCh38
          GenBankmRNA AA494170.1 - 1 94 Chr. 11 - 65426056 - 65426492 [+] on Build GRCh38
          GenBankmRNA AV692778.1 - 1 62 Chr. 11 - 65426056 - 65426383 [+] on Build GRCh38
          GenBankmRNA AV687822.1 - 80 Chr. - - [-] on Build
          GenBankmRNA BG573479.1 - 1 444 Chr. 11 - 65425674 - 65426375 [+] on Build GRCh38
          GenBankmRNA DA400808.1 - 1 357 Chr. 11 - 65425761 - 65426313 [+] on Build GRCh38
          GenBankmRNA BG620720.1 - 1 306 Chr. 11 - 65425812 - 65426513 [+] on Build GRCh38
          GenBankmRNA CB267546.1 - 1 295 Chr. 11 - 65425822 - 65426400 [+] on Build GRCh38
          GenBankmRNA AV661235.2 - 1 73 Chr. 11 - 65426045 - 65426395 [+] on Build GRCh38
          GenBankmRNA AA888363.1 - 1 77 Chr. 11 - 65426083 - 65426470 [+] on Build GRCh38
          GenBankmRNA BQ721690.1 - 1 518 Chr. 11 - 65425600 - 65426525 [+] on Build GRCh38
          GenBankmRNA BE931943.1 - 1 143 Chr. 11 - 65426007 - 65426385 [+] on Build GRCh38
          GenBankmRNA R71357.1 - 62 Chr. - - [-] on Build
          GenBankmRNA BE840488.1 - 1 88 Chr. 11 - 65426062 - 65426370 [+] on Build GRCh38
          GenBankmRNA CD000974.1 - 1 363 Chr. 11 - 65425757 - 65426413 [+] on Build GRCh38
          GenBankmRNA BX090547.1 - 1 31 Chr. 11 - 65426087 - 65426532 [+] on Build GRCh38
          GenBankmRNA AV686930.1 - 1 172 Chr. 11 - 65425946 - 65426357 [+] on Build GRCh38
          GenBankmRNA BE177867.1 - 1 119 Chr. 11 - 65426007 - 65426385 [+] on Build GRCh38
          GenBankmRNA CU448068.1 - 1 360 Chr. 11 - 65425767 - 65426285 [+] on Build GRCh38
          GenBankmRNA BE178390.1 - 1 118 Chr. 11 - 65426007 - 65426317 [+] on Build GRCh38
          GenBankmRNA BF882241.1 - 1 159 Chr. 11 - 65425964 - 65426424 [+] on Build GRCh38
          GenBankmRNA CU452644.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BF801704.1 - 1 306 Chr. 11 - 65425850 - 65426347 [+] on Build GRCh38
          GenBankmRNA AV654820.1 - 1 267 Chr. 11 - 65425851 - 65426187 [+] on Build GRCh38
          GenBankmRNA BF525951.1 - 1 186 Chr. 11 - 65425972 - 65426532 [+] on Build GRCh38
          GenBankmRNA CB958282.1 - 1 106 Chr. 11 - 65426013 - 65426525 [+] on Build GRCh38
          GenBankmRNA AV662136.2 - 1 84 Chr. 11 - 65426034 - 65426395 [+] on Build GRCh38
          GenBankmRNA AI525776.1 - 148 Chr. - - [-] on Build
          GenBankmRNA HX786077.1 - 1 529 Chr. 11 - 65425589 - 65426234 [+] on Build GRCh38
          GenBankmRNA EL955070.1 - 1 499 Chr. 11 - 65425621 - 65426180 [+] on Build GRCh38
          GenBankmRNA CB157506.1 - 1 307 Chr. 11 - 65425811 - 65426483 [+] on Build GRCh38
          GenBankmRNA CD674607.1 - 1 347 Chr. 11 - 65425771 - 65426353 [+] on Build GRCh38
          GenBankmRNA DA876302.1 - 1 162 Chr. 11 - 65425956 - 65426526 [+] on Build GRCh38
          GenBankmRNA BE178456.1 - 1 116 Chr. 11 - 65426016 - 65426385 [+] on Build GRCh38
          GenBankmRNA BE840565.1 - 1 88 Chr. 11 - 65426062 - 65426370 [+] on Build GRCh38
          GenBankmRNA CB107557.1 - 1 31 Chr. 11 - 65426087 - 65426496 [+] on Build GRCh38
          GenBankmRNA AA010989.1 - 1 323 Chr. 11 - 65425795 - 65426277 [+] on Build GRCh38
          GenBankmRNA AV654342.1 - 1 400 Chr. 11 - 65425718 - 65426385 [+] on Build GRCh38
          GenBankmRNA AV652583.1 - 1 299 Chr. 11 - 65425819 - 65426528 [+] on Build GRCh38
          GenBankmRNA BG113348.1 - 1 415 Chr. 11 - 65425699 - 65426357 [+] on Build GRCh38
          GenBankmRNA BM727960.1 - 1 127 Chr. 11 - 65425991 - 65426260 [+] on Build GRCh38
          GenBankmRNA BG674297.1 - 1 592 Chr. 11 - 65425524 - 65426351 [+] on Build GRCh38
          GenBankmRNA AW876895.1 - 1 185 Chr. 11 - 65425946 - 65426304 [+] on Build GRCh38
          GenBankmRNA HX787970.1 - 1 529 Chr. 11 - 65425589 - 65426234 [+] on Build GRCh38
          GenBankmRNA CD110553.1 - 540 Chr. - - [-] on Build
          GenBankmRNA BE842700.1 - 1 332 Chr. 11 - 65425796 - 65426304 [+] on Build GRCh38
          GenBankmRNA BU174238.1 - 1 530 Chr. 11 - 65425588 - 65426391 [+] on Build GRCh38
          GenBankmRNA AL691896.1 - 1 240 Chr. 11 - 65425879 - 65426528 [+] on Build GRCh38
          GenBankmRNA BM791400.1 - 1 238 Chr. 11 - 65425880 - 65426431 [+] on Build GRCh38
          GenBankmRNA CB962041.1 - 1 32 Chr. 11 - 65426088 - 65426532 [+] on Build GRCh38
          GenBankmRNA N23466.1 - 1 139 Chr. 11 - 65425979 - 65426413 [+] on Build GRCh38
          GenBankmRNA BX457383.2 - 1 734 Chr. 11 - 65425383 - 65426255 [+] on Build GRCh38
          GenBankmRNA W38857.1 - 168 Chr. - - [-] on Build
          GenBankmRNA CB963225.1 - 1 597 Chr. 11 - 65425523 - 65426287 [+] on Build GRCh38
          GenBankmRNA W56123.1 - 151 Chr. - - [-] on Build
          GenBankmRNA BE178393.1 - 1 114 Chr. 11 - 65426006 - 65426385 [+] on Build GRCh38
          GenBankmRNA AA345082.1 - 1 121 Chr. 11 - 65425997 - 65426402 [+] on Build GRCh38
          GenBankmRNA CB269074.1 - 1 319 Chr. 11 - 65425797 - 65426328 [+] on Build GRCh38
          GenBankmRNA CB161566.1 - 1 307 Chr. 11 - 65425811 - 65426354 [+] on Build GRCh38
          GenBankmRNA BM782508.1 - 1 387 Chr. 11 - 65425731 - 65426337 [+] on Build GRCh38
          GenBankmRNA BG341320.1 - 1 110 Chr. 11 - 65426009 - 65426416 [+] on Build GRCh38
          GenBankmRNA BY996298.1 - 1 501 Chr. 11 - 65425625 - 65426277 [+] on Build GRCh38
          GenBankmRNA BP423464.1 - 1 229 Chr. 11 - 65425889 - 65426398 [+] on Build GRCh38
          GenBankmRNA BG696830.1 - 1 649 Chr. 11 - 65425470 - 65426261 [+] on Build GRCh38
          GenBankmRNA AV733896.1 - 1 282 Chr. 11 - 65425836 - 65426525 [+] on Build GRCh38
          GenBankmRNA DV459908.1 - 1 32 Chr. 11 - 65426086 - 65426360 [+] on Build GRCh38
          GenBankmRNA AV697667.1 - 1 80 Chr. 11 - 65426038 - 65426471 [+] on Build GRCh38
          GenBankmRNA BE166954.1 - 1 151 Chr. 11 - 65425967 - 65426300 [+] on Build GRCh38
          GenBankmRNA BU664718.1 - 1 80 Chr. 11 - 65426043 - 65426630 [+] on Build GRCh38
          GenBankmRNA BE928878.1 - 1 289 Chr. 11 - 65425832 - 65426415 [+] on Build GRCh38
          GenBankmRNA AW023266.1 - 1 30 Chr. 11 - 65426094 - 65426429 [+] on Build GRCh38
          GenBankmRNA BE768457.1 - 1 223 Chr. 11 - 65425898 - 65426243 [+] on Build GRCh38
          GenBankmRNA CK005272.1 - 1 367 Chr. 11 - 65425753 - 65426390 [+] on Build GRCh38
          GenBankmRNA CB998941.1 - 1 341 Chr. 11 - 65425779 - 65426514 [+] on Build GRCh38
          GenBankmRNA BE840495.1 - 1 128 Chr. 11 - 65426016 - 65426231 [+] on Build GRCh38
          GenBankmRNA CU446474.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BP417015.1 - 1 440 Chr. 11 - 65425679 - 65426282 [+] on Build GRCh38
          GenBankmRNA BE834451.1 - 1 140 Chr. 11 - 65426007 - 65426385 [+] on Build GRCh38
          GenBankmRNA DR423495.1 - 1 598 Chr. 11 - 65425524 - 65426245 [+] on Build GRCh38
          GenBankmRNA BE840517.1 - 1 169 Chr. 11 - 65425963 - 65426370 [+] on Build GRCh38
          GenBankmRNA BE178394.1 - 1 112 Chr. 11 - 65426011 - 65426385 [+] on Build GRCh38
          GenBankmRNA CK430042.1 - 1 342 Chr. 11 - 65425776 - 65426354 [+] on Build GRCh38
          GenBankmRNA R22093.1 - 16 Chr. - - [-] on Build
          GenBankmRNA EL594402.1 - 1 296 Chr. 11 - 65425837 - 65426373 [+] on Build GRCh38
          GenBankmRNA BI832781.1 - 1 436 Chr. 11 - 65425674 - 65426408 [+] on Build GRCh38
          GenBankmRNA BM707319.1 - 1 254 Chr. 11 - 65425872 - 65426329 [+] on Build GRCh38
          GenBankmRNA W37277.1 - 114 Chr. - - [-] on Build
          GenBankmRNA AA578536.1 - 1 45 Chr. 11 - 65426073 - 65426363 [+] on Build GRCh38
          GenBankmRNA F35885.1 - 21 Chr. - - [-] on Build
          GenBankmRNA F25169.1 - 1 15 Chr. 11 - 65426103 - 65426508 [+] on Build GRCh38
          GenBankmRNA AA054357.1 - 1 61 Chr. 11 - 65426065 - 65426526 [+] on Build GRCh38
          GenBankmRNA BE840572.1 - 1 128 Chr. 11 - 65426016 - 65426231 [+] on Build GRCh38
          GenBankmRNA BF848999.1 - 248 Chr. - - [-] on Build
          GenBankmRNA BE825957.1 - 1 81 Chr. 11 - 65426044 - 65426420 [+] on Build GRCh38
          GenBankmRNA BQ694729.1 - 1 53 Chr. 11 - 65426065 - 65426522 [+] on Build GRCh38
          GenBankmRNA BE834414.1 - 1 139 Chr. 11 - 65426011 - 65426385 [+] on Build GRCh38
          GenBankmRNA CK430392.1 - 1 33 Chr. 11 - 65426085 - 65426522 [+] on Build GRCh38
          GenBankmRNA AA879050.1 - 1 15 Chr. 11 - 65426103 - 65426343 [+] on Build GRCh38
          GenBankmRNA AI459536.1 - 1 141 Chr. 11 - 65425977 - 65426406 [+] on Build GRCh38
          GenBankmRNA AA640879.1 - 1 45 Chr. 11 - 65426078 - 65426490 [+] on Build GRCh38
          GenBankmRNA BG674955.1 - 1 497 Chr. 11 - 65425622 - 65426434 [+] on Build GRCh38
          GenBankmRNA BE166789.1 - 1 183 Chr. 11 - 65425944 - 65426304 [+] on Build GRCh38
          GenBankmRNA BE166956.1 - 1 152 Chr. 11 - 65425966 - 65426304 [+] on Build GRCh38
          GenBankmRNA AA397478.1 - 1 45 Chr. 11 - 65426078 - 65426422 [+] on Build GRCh38
          GenBankmRNA AW999502.1 - 1 86 Chr. 11 - 65426055 - 65426376 [+] on Build GRCh38
          GenBankmRNA AA493725.1 - 1 105 Chr. 11 - 65426013 - 65426485 [+] on Build GRCh38
          GenBankmRNA AA569490.1 - 1 95 Chr. 11 - 65426027 - 65426428 [+] on Build GRCh38
          GenBankmRNA BE832339.1 - 1 114 Chr. 11 - 65426016 - 65426231 [+] on Build GRCh38
          GenBankmRNA CD721761.1 - 1 166 Chr. 11 - 65425952 - 65426524 [+] on Build GRCh38
          GenBankmRNA CU447522.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA CB134570.1 - 1 306 Chr. 11 - 65425812 - 65426376 [+] on Build GRCh38
          GenBankmRNA CU447848.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BU929248.1 - 1 209 Chr. 11 - 65425909 - 65426525 [+] on Build GRCh38
          GenBankmRNA BE834413.1 - 1 138 Chr. 11 - 65426007 - 65426385 [+] on Build GRCh38
          GenBankmRNA D58883.1 - 1 33 Chr. 11 - 65426085 - 65426363 [+] on Build GRCh38
          GenBankmRNA AA393236.1 - 1 166 Chr. 11 - 65425952 - 65426446 [+] on Build GRCh38
          GenBankmRNA CB155865.1 - 1 452 Chr. 11 - 65425666 - 65426262 [+] on Build GRCh38
          GenBankmRNA BG622240.1 - 159 Chr. - - [-] on Build
          GenBankmRNA BE840514.1 - 1 118 Chr. 11 - 65426011 - 65426231 [+] on Build GRCh38
          GenBankmRNA CU456991.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BQ721384.1 - 1 103 Chr. 11 - 65426054 - 65426526 [+] on Build GRCh38
          GenBankmRNA BE840692.1 - 1 118 Chr. 11 - 65426018 - 65426231 [+] on Build GRCh38
          GenBankmRNA CU443792.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA CB160909.1 - 1 223 Chr. 11 - 65425923 - 65426235 [+] on Build GRCh38
          GenBankmRNA CD722075.1 - 1 503 Chr. 11 - 65425615 - 65426202 [+] on Build GRCh38
          GenBankmRNA CB957193.1 - 1 17 Chr. 11 - 65426103 - 65426526 [+] on Build GRCh38
          GenBankmRNA DB277475.1 - 1 195 Chr. 11 - 65425923 - 65426402 [+] on Build GRCh38
          GenBankmRNA CB161416.1 - 1 307 Chr. 11 - 65425811 - 65426329 [+] on Build GRCh38
          GenBankmRNA BM756804.1 - 1 192 Chr. 11 - 65425925 - 65426512 [+] on Build GRCh38
          GenBankmRNA CU449328.1 - 1 360 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BQ695097.1 - 1 55 Chr. 11 - 65426063 - 65426525 [+] on Build GRCh38
          GenBankmRNA AV697669.1 - 1 80 Chr. 11 - 65426038 - 65426471 [+] on Build GRCh38
          GenBankmRNA BF361246.1 - 1 88 Chr. 11 - 65426033 - 65426385 [+] on Build GRCh38
          GenBankmRNA AA421498.1 - 1 16 Chr. 11 - 65426102 - 65426505 [+] on Build GRCh38
          GenBankmRNA CU445808.1 - 1 475 Chr. 11 - 65425654 - 65426228 [+] on Build GRCh38
          GenBankmRNA W38907.1 - 149 Chr. - - [-] on Build
          GenBankmRNA BE163870.1 - 1 206 Chr. 11 - 65425922 - 65426266 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0