Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110102.1 | 2 | 1845 | |||
GenBank mRNA | AK001533.1 | - | 2 | 1844 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN426486.1 | - | 2 | 393 | Chr. 7 - 99014617 - 99030978 [+] on Build GRCh38 |