Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100506159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104634.2 | 3 | 492 | |||
GenBank mRNA | BX339226.2 | - | 3 | 435 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA487642.1 | - | 3 | 443 | Chr. 12 - 9939923 - 9943447 [-] on Build GRCh38 |