Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HAGLR SIRNA Assays ›
          Gene Symbol:
          HAGLR
          Gene Name:
          HOXD antisense growth-associated long non-coding RNA
          Gene Aliases:
          HOXD-AS1, Mdgt
          Chromosome Location:
          Chr.2: 176173189 - 176188958 on Build GRCh38
          UniGene ID:
          Hs.98661
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n504307

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HAGLR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033979.2 3 365
          NR_110458.1 3 639
          NR_110459.1 5 590
          NR_110460.1 5 560
          NR_110461.1 4 495
          NR_110462.1 2 476
          NR_110463.1 4 458
          NR_110464.1 4 446
          NR_110465.1 4 444
          NR_110466.1 3 374
          NR_110467.1 3 347
          NR_110468.1 3 335
          GenBank mRNA BC030713.2 - 3 626
          BC047481.1 - 3 626
          DA398428.1 - 4 351
          DA736032.1 - 4 496
          DB045673.1 - 3 302
          DB100342.1 - 4 412
          DB100521.1 - 4 359
          HY018648.1 - 2 473
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA401217.1 - 2 294 Chr. 2 - 176176476 - 176177568 [-] on Build GRCh38
          GenBankmRNA DA331336.1 - 3 334 Chr. 2 - 176176386 - 176188531 [-] on Build GRCh38
          GenBankmRNA DA292668.1 - 3 310 Chr. 2 - 176176362 - 176188508 [-] on Build GRCh38
          GenBankmRNA DA727022.1 - 4 490 Chr. 2 - 176176553 - 176188264 [-] on Build GRCh38
          GenBankmRNA DA538103.1 - 3 342 Chr. 2 - 176176390 - 176188540 [-] on Build GRCh38
          GenBankmRNA DA755456.1 - 3 365 Chr. 2 - 176176460 - 176188958 [-] on Build GRCh38
          GenBankmRNA BG718435.1 - 3 623 Chr. 2 - 176176481 - 176178109 [-] on Build GRCh38
          GenBankmRNA BG717191.1 - 2 479 Chr. 2 - 176176472 - 176178125 [-] on Build GRCh38
          GenBankmRNA BE378592.1 - 4 368 Chr. 2 - 176176487 - 176179060 [-] on Build GRCh38
          GenBankmRNA BG324915.1 - 325 Chr. - - [-] on Build
          GenBankmRNA BI520150.1 - 3 629 Chr. 2 - 176176399 - 176178119 [-] on Build GRCh38
          GenBankmRNA DA182774.1 - 3 328 Chr. 2 - 176176390 - 176188526 [-] on Build GRCh38
          GenBankmRNA HY025996.1 - 3 307 Chr. 2 - 176164051 - 176178099 [-] on Build GRCh38
          GenBankmRNA AL120749.1 - 3 244 Chr. 2 - 176176169 - 176188442 [-] on Build GRCh38
          GenBankmRNA HY008350.1 - 3 324 Chr. 2 - 176176388 - 176178118 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0