Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097560.1 | - | 2 | 328 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB050922.1 | - | 2 | 251 | Chr. 15 - 69073003 - 69091759 [+] on Build GRCh38 |