Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700071M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045444.1 | 2, 3 | 1894 | |||
NR_045445.1 | 3, 4 | 350 | ||||
GenBank mRNA | AK133239.1 | - | 2, 3 | 1894 | ||
AK161361.1 | - | 3, 4 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN842847.1 | - | 346 | Chr. - - [-] on Build |