Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG5 SIRNA Assays ›
          Gene Symbol:
          Snhg5 9330159M07Rik 9430037G07Rik
          Gene Name:
          small nucleolar RNA host gene 5
          RIKEN cDNA 9330159M07 gene
          RIKEN cDNA 9430037G07 gene
          Gene Aliases:
          2810026P18Rik, 4933428I09Rik, mU50HG-b
          1700096K11Rik, mU50hg-a
          -
          Chromosome Location:
          Chr.9: 88521052 - 88522897 on Build GRCm38
          Chromosome Location:
          Chr.9: 88841798 - 88858870 on Build GRCm38
          Chromosome Location:
          Chr.9: 88595325 - 88599243 on Build GRCm38
          UniGene ID:
          Mm.21798
          Mm.149040
          Mm.427378
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID n394357

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Snhg5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_040721.1 5 814
          GenBank mRNA AK012825.1 - 5 806
          BC032970.1 - 5 789
          KF876690.1 - 5 808
          Gene Symbol: 9330159M07Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK136619.1 - 5 420
          Gene Symbol: 9430037G07Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_040766.1 5 465
          GenBank mRNA AK034782.1 - 3 1027
          AK050253.1 - 5 466
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA231154.1 - 4 347 Chr. 9 - 88521097 - 88522328 [-] on Build GRCm38
          GenBankmRNA AV445958.1 - 3 315 Chr. 9 - 88521083 - 88521965 [-] on Build GRCm38
          GenBankmRNA CA539769.1 - 5 516 Chr. 9 - 88521162 - 88522587 [-] on Build GRCm38
          GenBankmRNA CB194977.1 - 6 532 Chr. 9 - 88521064 - 88522879 [-] on Build GRCm38
          GenBankmRNA CA983870.1 - 5 796 Chr. 9 - 88521105 - 88522878 [-] on Build GRCm38
          GenBankmRNA CA979224.1 - 5 779 Chr. 9 - 88521143 - 88522862 [-] on Build GRCm38
          GenBankmRNA BG802512.1 - 5 499 Chr. 9 - 88521233 - 88522582 [-] on Build GRCm38
          GenBankmRNA BE646905.1 - 3 403 Chr. 9 - 88521148 - 88522053 [-] on Build GRCm38
          GenBankmRNA CD563600.1 - 5 496 Chr. 9 - 88521139 - 88522579 [-] on Build GRCm38
          GenBankmRNA BI440435.1 - 6 522 Chr. 9 - 88521221 - 88522866 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
          git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0