Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EBLN3 SIRNA Assays ›
          Gene Symbol:
          EBLN3 ZCCHC7
          Gene Name:
          endogenous Bornavirus-like nucleoprotein 3
          zinc finger CCHC-type containing 7
          Gene Aliases:
          -
          AIR1, HSPC086
          Chromosome Location:
          Chr.9: 37079896 - 37090401 on Build GRCh38
          Chromosome Location:
          Chr.9: 37120169 - 37358149 on Build GRCh38
          UniGene ID:
          Hs.446271
          Hs.654700
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n360604

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EBLN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036592.1 1 461
          GenBank mRNA AK091718.1 - 1 461
          BC032471.1 - 1 360
          BC033624.1 - 1 360
          BC080614.1 - 1 331
          Gene Symbol: ZCCHC7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005251608.4 XP_005251665.1 1 337
          XM_017015198.1 XP_016870687.1 1 36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ082270.1 - 1 119 Chr. 9 - 37080238 - 37086745 [+] on Build GRCh38
          GenBankmRNA CN360591.1 - 1 423 Chr. 9 - 37079957 - 37086835 [+] on Build GRCh38
          GenBankmRNA HY116837.1 - 1 34 Chr. 9 - 37080324 - 37086929 [+] on Build GRCh38
          GenBankmRNA AL560171.3 - 1 429 Chr. 9 - 37079928 - 37086741 [+] on Build GRCh38
          GenBankmRNA BI261355.1 - 1 331 Chr. 9 - 37080026 - 37086888 [+] on Build GRCh38
          GenBankmRNA BG257880.1 - 1 356 Chr. 9 - 37080011 - 37086830 [+] on Build GRCh38
          GenBankmRNA HY118934.1 - 1 203 Chr. 9 - 37080154 - 37086752 [+] on Build GRCh38
          GenBankmRNA BX443970.2 - 1 370 Chr. 9 - 37079986 - 37086901 [+] on Build GRCh38
          GenBankmRNA BM013417.1 - 1 381 Chr. 9 - 37079976 - 37086898 [+] on Build GRCh38
          GenBankmRNA BM804156.1 - 312 Chr. - - [-] on Build
          GenBankmRNA BQ008189.1 - 1 412 Chr. 9 - 37079958 - 37086835 [+] on Build GRCh38
          GenBankmRNA BQ064157.1 - 1 329 Chr. 9 - 37080028 - 37087062 [+] on Build GRCh38
          GenBankmRNA BM927474.1 - 1 30 Chr. 9 - 37080327 - 37087414 [+] on Build GRCh38
          GenBankmRNA HY092644.1 - 2 135 Chr. 9 - 37079915 - 37086757 [+] on Build GRCh38
          GenBankmRNA BI832820.1 - 1 360 Chr. 9 - 37079997 - 37087017 [+] on Build GRCh38
          GenBankmRNA BU931386.1 - 1 392 Chr. 9 - 37080001 - 37086976 [+] on Build GRCh38
          GenBankmRNA BM555131.1 - 320 Chr. - - [-] on Build
          GenBankmRNA BX324344.2 - 1 360 Chr. 9 - 37080000 - 37086787 [+] on Build GRCh38
          GenBankmRNA CN360594.1 - 1 384 Chr. 9 - 37079973 - 37086779 [+] on Build GRCh38
          GenBankmRNA HY130506.1 - 1 193 Chr. 9 - 37080165 - 37086780 [+] on Build GRCh38
          GenBankmRNA BG470765.1 - 1 422 Chr. 9 - 37079944 - 37086857 [+] on Build GRCh38
          GenBankmRNA DA074751.1 - 1 342 Chr. 9 - 37080015 - 37086711 [+] on Build GRCh38
          GenBankmRNA HY133719.1 - 1 203 Chr. 9 - 37080154 - 37086744 [+] on Build GRCh38
          GenBankmRNA BE397689.1 - 1 405 Chr. 9 - 37079991 - 37086753 [+] on Build GRCh38
          GenBankmRNA CN360593.1 - 1 363 Chr. 9 - 37079997 - 37086844 [+] on Build GRCh38
          GenBankmRNA BQ055290.1 - 1 30 Chr. 9 - 37080327 - 37087345 [+] on Build GRCh38
          GenBankmRNA AA448021.1 - 1 44 Chr. 9 - 37080312 - 37086801 [+] on Build GRCh38
          GenBankmRNA DR421805.1 - 1 405 Chr. 9 - 37079952 - 37086804 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding
          zinc ion binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0