Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100131840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057355.1 | - | 1 | 915 |
Gene Symbol: CCDC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713124.3 | XP_006713187.1 | 1 | 78 | ||
XM_011533673.2 | XP_011531975.1 | 1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB447408.1 | - | 1 | 82 | Chr. 3 - 49199617 - 49236574 [+] on Build GRCh38 |