Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEAT1 SIRNA Assays ›
          Gene Symbol:
          NEAT1
          Gene Name:
          nuclear paraspeckle assembly transcript 1 (non-protein coding)
          Gene Aliases:
          LINC00084, NCRNA00084, TncRNA, VINC
          Chromosome Location:
          Chr.11: 65422798 - 65445540 on Build GRCh38
          UniGene ID:
          Hs.523789
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272463

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEAT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028272.1 1 3095
          NR_131012.1 1 3095
          GenBank mRNA AF001893.1 - 1 847
          AK027191.1 - 1 1079
          EF177379.1 - 1 3095
          GQ859162.1 - 1 3095
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX457383.2 - 1 509 Chr. 11 - 65425383 - 65426255 [+] on Build GRCh38
          GenBankmRNA CB963225.1 - 1 372 Chr. 11 - 65425523 - 65426287 [+] on Build GRCh38
          GenBankmRNA CB269074.1 - 1 95 Chr. 11 - 65425797 - 65426328 [+] on Build GRCh38
          GenBankmRNA BM782508.1 - 1 162 Chr. 11 - 65425731 - 65426337 [+] on Build GRCh38
          GenBankmRNA CB161566.1 - 1 82 Chr. 11 - 65425811 - 65426354 [+] on Build GRCh38
          GenBankmRNA BY996298.1 - 1 276 Chr. 11 - 65425625 - 65426277 [+] on Build GRCh38
          GenBankmRNA AV733896.1 - 1 57 Chr. 11 - 65425836 - 65426525 [+] on Build GRCh38
          GenBankmRNA BG696830.1 - 1 424 Chr. 11 - 65425470 - 65426261 [+] on Build GRCh38
          GenBankmRNA BM796270.1 - 1 13 Chr. 11 - 65425880 - 65426480 [+] on Build GRCh38
          GenBankmRNA CB854402.1 - 1 318 Chr. 11 - 65425586 - 65426184 [+] on Build GRCh38
          GenBankmRNA AJ710291.1 - 1 167 Chr. 11 - 65425737 - 65426279 [+] on Build GRCh38
          GenBankmRNA BE928878.1 - 1 65 Chr. 11 - 65425832 - 65426415 [+] on Build GRCh38
          GenBankmRNA CK005272.1 - 1 142 Chr. 11 - 65425753 - 65426390 [+] on Build GRCh38
          GenBankmRNA CB998941.1 - 1 116 Chr. 11 - 65425779 - 65426514 [+] on Build GRCh38
          GenBankmRNA CU446474.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BP417015.1 - 1 215 Chr. 11 - 65425679 - 65426282 [+] on Build GRCh38
          GenBankmRNA DR423495.1 - 1 373 Chr. 11 - 65425524 - 65426245 [+] on Build GRCh38
          GenBankmRNA CD240095.1 - 1 160 Chr. 11 - 65425784 - 65426507 [+] on Build GRCh38
          GenBankmRNA BF084880.1 - 1 51 Chr. 11 - 65425844 - 65426432 [+] on Build GRCh38
          GenBankmRNA CB162181.1 - 1 64 Chr. 11 - 65425829 - 65426354 [+] on Build GRCh38
          GenBankmRNA BE928827.1 - 1 49 Chr. 11 - 65425844 - 65426390 [+] on Build GRCh38
          GenBankmRNA CK430042.1 - 1 117 Chr. 11 - 65425776 - 65426354 [+] on Build GRCh38
          GenBankmRNA EL594402.1 - 1 71 Chr. 11 - 65425837 - 65426373 [+] on Build GRCh38
          GenBankmRNA AL547749.3 - 1 379 Chr. 11 - 65425515 - 65426391 [+] on Build GRCh38
          GenBankmRNA BM707319.1 - 1 29 Chr. 11 - 65425872 - 65426329 [+] on Build GRCh38
          GenBankmRNA BE842714.1 - 1 126 Chr. 11 - 65425773 - 65426304 [+] on Build GRCh38
          GenBankmRNA BG740944.1 - 1 375 Chr. 11 - 65425517 - 65426312 [+] on Build GRCh38
          GenBankmRNA BG573479.1 - 1 219 Chr. 11 - 65425674 - 65426375 [+] on Build GRCh38
          GenBankmRNA BG621344.1 - 1 304 Chr. 11 - 65425589 - 65426359 [+] on Build GRCh38
          GenBankmRNA DA400808.1 - 1 132 Chr. 11 - 65425761 - 65426313 [+] on Build GRCh38
          GenBankmRNA BF679881.1 - 1 184 Chr. 11 - 65425708 - 65426288 [+] on Build GRCh38
          GenBankmRNA BG620720.1 - 1 81 Chr. 11 - 65425812 - 65426513 [+] on Build GRCh38
          GenBankmRNA CB267546.1 - 1 71 Chr. 11 - 65425822 - 65426400 [+] on Build GRCh38
          GenBankmRNA BQ721690.1 - 1 293 Chr. 11 - 65425600 - 65426525 [+] on Build GRCh38
          GenBankmRNA BG674955.1 - 1 272 Chr. 11 - 65425622 - 65426434 [+] on Build GRCh38
          GenBankmRNA CD000974.1 - 1 138 Chr. 11 - 65425757 - 65426413 [+] on Build GRCh38
          GenBankmRNA CU448068.1 - 1 135 Chr. 11 - 65425767 - 65426285 [+] on Build GRCh38
          GenBankmRNA AV654820.1 - 1 42 Chr. 11 - 65425851 - 65426187 [+] on Build GRCh38
          GenBankmRNA CU452644.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BF801704.1 - 1 81 Chr. 11 - 65425850 - 65426347 [+] on Build GRCh38
          GenBankmRNA CU447522.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA CB134570.1 - 1 81 Chr. 11 - 65425812 - 65426376 [+] on Build GRCh38
          GenBankmRNA CU447848.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA HX786077.1 - 1 304 Chr. 11 - 65425589 - 65426234 [+] on Build GRCh38
          GenBankmRNA BF084879.1 - 1 64 Chr. 11 - 65425836 - 65426429 [+] on Build GRCh38
          GenBankmRNA EL955070.1 - 1 274 Chr. 11 - 65425621 - 65426180 [+] on Build GRCh38
          GenBankmRNA CB157506.1 - 1 82 Chr. 11 - 65425811 - 65426483 [+] on Build GRCh38
          GenBankmRNA CD674607.1 - 1 122 Chr. 11 - 65425771 - 65426353 [+] on Build GRCh38
          GenBankmRNA CB155865.1 - 1 227 Chr. 11 - 65425666 - 65426262 [+] on Build GRCh38
          GenBankmRNA CU456991.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA AA010989.1 - 1 98 Chr. 11 - 65425795 - 65426277 [+] on Build GRCh38
          GenBankmRNA AV654342.1 - 1 175 Chr. 11 - 65425718 - 65426385 [+] on Build GRCh38
          GenBankmRNA AV652583.1 - 1 74 Chr. 11 - 65425819 - 65426528 [+] on Build GRCh38
          GenBankmRNA CD722075.1 - 1 278 Chr. 11 - 65425615 - 65426202 [+] on Build GRCh38
          GenBankmRNA CU443792.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA BG113348.1 - 1 192 Chr. 11 - 65425699 - 65426357 [+] on Build GRCh38
          GenBankmRNA CB161416.1 - 1 82 Chr. 11 - 65425811 - 65426329 [+] on Build GRCh38
          GenBankmRNA CU449328.1 - 1 135 Chr. 11 - 65425767 - 65426431 [+] on Build GRCh38
          GenBankmRNA HX787970.1 - 1 304 Chr. 11 - 65425589 - 65426234 [+] on Build GRCh38
          GenBankmRNA BE842700.1 - 1 107 Chr. 11 - 65425796 - 65426304 [+] on Build GRCh38
          GenBankmRNA CU445808.1 - 1 250 Chr. 11 - 65425654 - 65426228 [+] on Build GRCh38
          GenBankmRNA BU174238.1 - 1 305 Chr. 11 - 65425588 - 65426391 [+] on Build GRCh38
          GenBankmRNA AL691896.1 - 1 15 Chr. 11 - 65425879 - 65426528 [+] on Build GRCh38
          GenBankmRNA BM791400.1 - 1 13 Chr. 11 - 65425880 - 65426431 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0