Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TTC39C SIRNA Assays ›
          Gene Symbol:
          TTC39C
          Gene Name:
          tetratricopeptide repeat domain 39C
          Gene Aliases:
          C18orf17, HsT2697
          Chromosome Location:
          Chr.18: 23992773 - 24135952 on Build GRCh38
          UniGene ID:
          Hs.128576
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270646

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TTC39C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243425.1 NP_001230354.1 2 677
          XM_005258198.4 XP_005258255.1 2 160
          XM_017025550.1 XP_016881039.1 2 254
          GenBank mRNA AK074875.1 - 2 299
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE465341.1 - 2 204 Chr. 18 - 24014893 - 24020288 [+] on Build GRCh38
          GenBankmRNA BF197725.1 - 2 206 Chr. 18 - 24014892 - 24020183 [+] on Build GRCh38
          GenBankmRNA AW612284.1 - 2 184 Chr. 18 - 24014913 - 24019987 [+] on Build GRCh38
          GenBankmRNA BF477582.1 - 2 133 Chr. 18 - 24014964 - 24020255 [+] on Build GRCh38
          GenBankmRNA AI681193.1 - 2 205 Chr. 18 - 24014892 - 24020218 [+] on Build GRCh38
          GenBankmRNA AW138334.1 - 2 204 Chr. 18 - 24014893 - 24020012 [+] on Build GRCh38
          GenBankmRNA AI523823.1 - 2 184 Chr. 18 - 24014913 - 24020227 [+] on Build GRCh38
          GenBankmRNA AW470336.1 - 2 184 Chr. 18 - 24014916 - 24020076 [+] on Build GRCh38
          GenBankmRNA AI632691.1 - 2 184 Chr. 18 - 24014913 - 24020080 [+] on Build GRCh38
          GenBankmRNA AA972636.1 - 2 200 Chr. 18 - 24014892 - 24020152 [+] on Build GRCh38
          GenBankmRNA AW139596.1 - 2 204 Chr. 18 - 24014893 - 24020231 [+] on Build GRCh38
          GenBankmRNA BF058122.1 - 2 205 Chr. 18 - 24014892 - 24020182 [+] on Build GRCh38
          GenBankmRNA W72622.1 - 201 Chr. - - [-] on Build
          GenBankmRNA AW206969.1 - 2 204 Chr. 18 - 24014893 - 24020112 [+] on Build GRCh38
          GenBankmRNA AI985561.1 - 2 184 Chr. 18 - 24014913 - 24020198 [+] on Build GRCh38
          GenBankmRNA AW149586.1 - 2 184 Chr. 18 - 24014913 - 24020225 [+] on Build GRCh38
          GenBankmRNA AA806974.1 - 2 126 Chr. 18 - 24014971 - 24020221 [+] on Build GRCh38
          GenBankmRNA AA987961.1 - 2 183 Chr. 18 - 24014909 - 24020231 [+] on Build GRCh38
          GenBankmRNA AA947936.1 - 2 178 Chr. 18 - 24014917 - 24020222 [+] on Build GRCh38
          GenBankmRNA AW015583.1 - 2 204 Chr. 18 - 24014893 - 24020197 [+] on Build GRCh38
          GenBankmRNA BE327092.1 - 2 205 Chr. 18 - 24014892 - 24020266 [+] on Build GRCh38
          GenBankmRNA BE673679.1 - 2 206 Chr. 18 - 24014892 - 24020254 [+] on Build GRCh38
          GenBankmRNA BE326333.1 - 2 205 Chr. 18 - 24014892 - 24020195 [+] on Build GRCh38
          GenBankmRNA AW138149.1 - 2 204 Chr. 18 - 24014893 - 24020105 [+] on Build GRCh38
          GenBankmRNA AI701287.1 - 2 185 Chr. 18 - 24014916 - 24019993 [+] on Build GRCh38
          GenBankmRNA BE219161.1 - 2 205 Chr. 18 - 24014892 - 24020226 [+] on Build GRCh38
          GenBankmRNA AI693271.1 - 2 162 Chr. 18 - 24014936 - 24020208 [+] on Build GRCh38
          GenBankmRNA BF057123.1 - 2 205 Chr. 18 - 24014892 - 24020196 [+] on Build GRCh38
          GenBankmRNA BG149174.1 - 2 159 Chr. 18 - 24014938 - 24020213 [+] on Build GRCh38
          GenBankmRNA AI334306.1 - 2 184 Chr. 18 - 24014913 - 24020211 [+] on Build GRCh38
          GenBankmRNA AA826395.1 - 2 153 Chr. 18 - 24014943 - 24020118 [+] on Build GRCh38
          GenBankmRNA BF108457.1 - 2 206 Chr. 18 - 24014892 - 24020141 [+] on Build GRCh38
          GenBankmRNA AI696060.1 - 2 205 Chr. 18 - 24014892 - 24020187 [+] on Build GRCh38
          GenBankmRNA HY051839.1 - 2 334 Chr. 18 - 24014763 - 24020000 [+] on Build GRCh38
          GenBankmRNA AW052123.1 - 2 183 Chr. 18 - 24014914 - 24020192 [+] on Build GRCh38
          GenBankmRNA AI298733.1 - 2 184 Chr. 18 - 24014913 - 24020203 [+] on Build GRCh38
          GenBankmRNA AI767968.1 - 2 184 Chr. 18 - 24014913 - 24020218 [+] on Build GRCh38
          GenBankmRNA AI475742.1 - 2 184 Chr. 18 - 24014913 - 24020202 [+] on Build GRCh38
          GenBankmRNA AW167659.1 - 2 184 Chr. 18 - 24014913 - 24020177 [+] on Build GRCh38
          GenBankmRNA AI350132.1 - 2 184 Chr. 18 - 24014913 - 24020166 [+] on Build GRCh38
          GenBankmRNA BE467775.1 - 2 187 Chr. 18 - 24014910 - 24020083 [+] on Build GRCh38
          GenBankmRNA AA488904.1 - 2 132 Chr. 18 - 24014968 - 24020108 [+] on Build GRCh38
          GenBankmRNA BE328182.1 - 2 187 Chr. 18 - 24014910 - 24020079 [+] on Build GRCh38
          GenBankmRNA BE674575.1 - 2 206 Chr. 18 - 24014892 - 24020195 [+] on Build GRCh38
          GenBankmRNA AW968939.1 - 2 188 Chr. 18 - 24014909 - 24020259 [+] on Build GRCh38
          GenBankmRNA AA410446.1 - 2 205 Chr. 18 - 24014892 - 24020160 [+] on Build GRCh38
          GenBankmRNA HY026865.1 - 2 460 Chr. 18 - 24014637 - 24019941 [+] on Build GRCh38
          GenBankmRNA BE217897.1 - 2 205 Chr. 18 - 24014892 - 24020255 [+] on Build GRCh38
          GenBankmRNA BF433075.1 - 2 206 Chr. 18 - 24014892 - 24020285 [+] on Build GRCh38
          GenBankmRNA AI801957.1 - 2 205 Chr. 18 - 24014892 - 24020229 [+] on Build GRCh38
          GenBankmRNA BM701431.1 - 2 171 Chr. 18 - 24014926 - 24020187 [+] on Build GRCh38
          GenBankmRNA BG150505.1 - 2 185 Chr. 18 - 24014912 - 24020058 [+] on Build GRCh38
          GenBankmRNA BE222387.1 - 2 203 Chr. 18 - 24014894 - 24020083 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0