Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TTC39C SIRNA Assays ›
          Gene Symbol:
          TTC39C
          Gene Name:
          tetratricopeptide repeat domain 39C
          Gene Aliases:
          C18orf17, HsT2697
          Chromosome Location:
          Chr.18: 23992773 - 24135952 on Build GRCh38
          UniGene ID:
          Hs.128576
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270644

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TTC39C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243425.1 NP_001230354.1 2 737
          XM_005258198.4 XP_005258255.1 2 220
          XM_017025550.1 XP_016881039.1 2 314
          GenBank mRNA AK074875.1 - 2 359
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE465341.1 - 2 264 Chr. 18 - 24014893 - 24020288 [+] on Build GRCh38
          GenBankmRNA BF197725.1 - 2 266 Chr. 18 - 24014892 - 24020183 [+] on Build GRCh38
          GenBankmRNA BF477582.1 - 2 193 Chr. 18 - 24014964 - 24020255 [+] on Build GRCh38
          GenBankmRNA AW612284.1 - 2 244 Chr. 18 - 24014913 - 24019987 [+] on Build GRCh38
          GenBankmRNA AW138334.1 - 2 264 Chr. 18 - 24014893 - 24020012 [+] on Build GRCh38
          GenBankmRNA AI681193.1 - 2 265 Chr. 18 - 24014892 - 24020218 [+] on Build GRCh38
          GenBankmRNA AI523823.1 - 2 244 Chr. 18 - 24014913 - 24020227 [+] on Build GRCh38
          GenBankmRNA AW470336.1 - 2 244 Chr. 18 - 24014916 - 24020076 [+] on Build GRCh38
          GenBankmRNA AI632691.1 - 2 244 Chr. 18 - 24014913 - 24020080 [+] on Build GRCh38
          GenBankmRNA AA972636.1 - 2 260 Chr. 18 - 24014892 - 24020152 [+] on Build GRCh38
          GenBankmRNA AW139596.1 - 2 264 Chr. 18 - 24014893 - 24020231 [+] on Build GRCh38
          GenBankmRNA BF058122.1 - 2 265 Chr. 18 - 24014892 - 24020182 [+] on Build GRCh38
          GenBankmRNA W72622.1 - 261 Chr. - - [-] on Build
          GenBankmRNA AW206969.1 - 2 264 Chr. 18 - 24014893 - 24020112 [+] on Build GRCh38
          GenBankmRNA AI985561.1 - 2 244 Chr. 18 - 24014913 - 24020198 [+] on Build GRCh38
          GenBankmRNA AA806974.1 - 2 186 Chr. 18 - 24014971 - 24020221 [+] on Build GRCh38
          GenBankmRNA AA987961.1 - 2 243 Chr. 18 - 24014909 - 24020231 [+] on Build GRCh38
          GenBankmRNA AA947936.1 - 2 238 Chr. 18 - 24014917 - 24020222 [+] on Build GRCh38
          GenBankmRNA AW015583.1 - 2 264 Chr. 18 - 24014893 - 24020197 [+] on Build GRCh38
          GenBankmRNA BE327092.1 - 2 265 Chr. 18 - 24014892 - 24020266 [+] on Build GRCh38
          GenBankmRNA BE673679.1 - 2 266 Chr. 18 - 24014892 - 24020254 [+] on Build GRCh38
          GenBankmRNA BE326333.1 - 2 265 Chr. 18 - 24014892 - 24020195 [+] on Build GRCh38
          GenBankmRNA AW138149.1 - 2 264 Chr. 18 - 24014893 - 24020105 [+] on Build GRCh38
          GenBankmRNA BE219161.1 - 2 265 Chr. 18 - 24014892 - 24020226 [+] on Build GRCh38
          GenBankmRNA AI693271.1 - 2 222 Chr. 18 - 24014936 - 24020208 [+] on Build GRCh38
          GenBankmRNA BF057123.1 - 2 265 Chr. 18 - 24014892 - 24020196 [+] on Build GRCh38
          GenBankmRNA BG149174.1 - 2 219 Chr. 18 - 24014938 - 24020213 [+] on Build GRCh38
          GenBankmRNA AI334306.1 - 2 244 Chr. 18 - 24014913 - 24020211 [+] on Build GRCh38
          GenBankmRNA AA826395.1 - 2 213 Chr. 18 - 24014943 - 24020118 [+] on Build GRCh38
          GenBankmRNA BF108457.1 - 2 266 Chr. 18 - 24014892 - 24020141 [+] on Build GRCh38
          GenBankmRNA AI696060.1 - 2 265 Chr. 18 - 24014892 - 24020187 [+] on Build GRCh38
          GenBankmRNA HY051839.1 - 2 394 Chr. 18 - 24014763 - 24020000 [+] on Build GRCh38
          GenBankmRNA AW052123.1 - 2 243 Chr. 18 - 24014914 - 24020192 [+] on Build GRCh38
          GenBankmRNA AI298733.1 - 2 244 Chr. 18 - 24014913 - 24020203 [+] on Build GRCh38
          GenBankmRNA AI767968.1 - 2 244 Chr. 18 - 24014913 - 24020218 [+] on Build GRCh38
          GenBankmRNA AI475742.1 - 2 244 Chr. 18 - 24014913 - 24020202 [+] on Build GRCh38
          GenBankmRNA AW167659.1 - 2 244 Chr. 18 - 24014913 - 24020177 [+] on Build GRCh38
          GenBankmRNA AI350132.1 - 2 241 Chr. 18 - 24014913 - 24020166 [+] on Build GRCh38
          GenBankmRNA BE467775.1 - 2 247 Chr. 18 - 24014910 - 24020083 [+] on Build GRCh38
          GenBankmRNA AA488904.1 - 2 192 Chr. 18 - 24014968 - 24020108 [+] on Build GRCh38
          GenBankmRNA BE328182.1 - 2 247 Chr. 18 - 24014910 - 24020079 [+] on Build GRCh38
          GenBankmRNA BE674575.1 - 2 266 Chr. 18 - 24014892 - 24020195 [+] on Build GRCh38
          GenBankmRNA AA410446.1 - 2 265 Chr. 18 - 24014892 - 24020160 [+] on Build GRCh38
          GenBankmRNA AW968939.1 - 2 248 Chr. 18 - 24014909 - 24020259 [+] on Build GRCh38
          GenBankmRNA BE217897.1 - 2 265 Chr. 18 - 24014892 - 24020255 [+] on Build GRCh38
          GenBankmRNA BF433075.1 - 2 266 Chr. 18 - 24014892 - 24020285 [+] on Build GRCh38
          GenBankmRNA AI801957.1 - 2 265 Chr. 18 - 24014892 - 24020229 [+] on Build GRCh38
          GenBankmRNA BM701431.1 - 2 231 Chr. 18 - 24014926 - 24020187 [+] on Build GRCh38
          GenBankmRNA BG150505.1 - 2 245 Chr. 18 - 24014912 - 24020058 [+] on Build GRCh38
          GenBankmRNA DA734643.1 - 2 359 Chr. 18 - 24014799 - 24020073 [+] on Build GRCh38
          GenBankmRNA BE222387.1 - 2 263 Chr. 18 - 24014894 - 24020083 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0