Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TTC39C SIRNA Assays ›
          Gene Symbol:
          TTC39C
          Gene Name:
          tetratricopeptide repeat domain 39C
          Gene Aliases:
          C18orf17, HsT2697
          Chromosome Location:
          Chr.18: 23992773 - 24135952 on Build GRCh38
          UniGene ID:
          Hs.128576
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270643

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TTC39C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243425.1 NP_001230354.1 2 936
          GenBank mRNA AK074875.1 - 2 558
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AI334306.1 - 2 443 Chr. 18 - 24014913 - 24020211 [+] on Build GRCh38
          GenBankmRNA AW149586.1 - 2 445 Chr. 18 - 24014913 - 24020225 [+] on Build GRCh38
          GenBankmRNA BE465341.1 - 2 463 Chr. 18 - 24014893 - 24020288 [+] on Build GRCh38
          GenBankmRNA AA806974.1 - 2 385 Chr. 18 - 24014971 - 24020221 [+] on Build GRCh38
          GenBankmRNA BF477582.1 - 2 392 Chr. 18 - 24014964 - 24020255 [+] on Build GRCh38
          GenBankmRNA BE674575.1 - 2 465 Chr. 18 - 24014892 - 24020195 [+] on Build GRCh38
          GenBankmRNA AI523823.1 - 2 443 Chr. 18 - 24014913 - 24020227 [+] on Build GRCh38
          GenBankmRNA AA987961.1 - 2 442 Chr. 18 - 24014909 - 24020231 [+] on Build GRCh38
          GenBankmRNA AA947936.1 - 2 437 Chr. 18 - 24014917 - 24020222 [+] on Build GRCh38
          GenBankmRNA AW015583.1 - 2 463 Chr. 18 - 24014893 - 24020197 [+] on Build GRCh38
          GenBankmRNA AW968939.1 - 2 447 Chr. 18 - 24014909 - 24020259 [+] on Build GRCh38
          GenBankmRNA BE326333.1 - 2 464 Chr. 18 - 24014892 - 24020195 [+] on Build GRCh38
          GenBankmRNA BF433075.1 - 2 465 Chr. 18 - 24014892 - 24020285 [+] on Build GRCh38
          GenBankmRNA BE217897.1 - 2 464 Chr. 18 - 24014892 - 24020255 [+] on Build GRCh38
          GenBankmRNA AW052123.1 - 2 442 Chr. 18 - 24014914 - 24020192 [+] on Build GRCh38
          GenBankmRNA AW139596.1 - 2 463 Chr. 18 - 24014893 - 24020231 [+] on Build GRCh38
          GenBankmRNA AI693271.1 - 2 421 Chr. 18 - 24014936 - 24020208 [+] on Build GRCh38
          GenBankmRNA AI298733.1 - 2 443 Chr. 18 - 24014913 - 24020203 [+] on Build GRCh38
          GenBankmRNA BF057123.1 - 2 464 Chr. 18 - 24014892 - 24020196 [+] on Build GRCh38
          GenBankmRNA AI767968.1 - 2 443 Chr. 18 - 24014913 - 24020218 [+] on Build GRCh38
          GenBankmRNA AI475742.1 - 2 443 Chr. 18 - 24014913 - 24020202 [+] on Build GRCh38
          GenBankmRNA BG149174.1 - 2 418 Chr. 18 - 24014938 - 24020213 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0