Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC01089 SIRNA Assays ›
          Gene Symbol:
          LINC01089
          Gene Name:
          long intergenic non-protein coding RNA 1089
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 121795267 - 121803484 on Build GRCh38
          UniGene ID:
          Hs.524804
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n262580

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC01089
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002809.2 2 692
          GenBank mRNA AK311480.1 - 1 248
          BC038786.1 - 2 692
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA142385.1 - 1 247 Chr. 12 - 121799869 - 121802945 [-] on Build GRCh38
          GenBankmRNA BI818784.1 - 2 141 Chr. 12 - 121795804 - 121802933 [-] on Build GRCh38
          GenBankmRNA BG035222.1 - 193 Chr. - - [-] on Build
          GenBankmRNA HY093841.1 - 1 248 Chr. 12 - 121799882 - 121802945 [-] on Build GRCh38
          GenBankmRNA DC377812.1 - 1 248 Chr. 12 - 121799622 - 121802946 [-] on Build GRCh38
          GenBankmRNA Z43897.1 - 141 Chr. - - [-] on Build
          GenBankmRNA BI822154.1 - 2 141 Chr. 12 - 121802074 - 121802933 [-] on Build GRCh38
          GenBankmRNA BM470301.1 - 134 Chr. - - [-] on Build
          GenBankmRNA CN310935.1 - 1 187 Chr. 12 - 121799757 - 121802883 [-] on Build GRCh38
          GenBankmRNA BM544353.1 - 162 Chr. - - [-] on Build
          GenBankmRNA CN310936.1 - 1 224 Chr. 12 - 121796221 - 121802920 [-] on Build GRCh38
          GenBankmRNA CD516035.1 - 2 128 Chr. 12 - 121795815 - 121802920 [-] on Build GRCh38
          GenBankmRNA DA925861.1 - 2 153 Chr. 12 - 121802279 - 121802945 [-] on Build GRCh38
          GenBankmRNA CN398873.1 - 2 150 Chr. 12 - 121795590 - 121802930 [-] on Build GRCh38
          GenBankmRNA BM698927.1 - 1 222 Chr. 12 - 121796211 - 121802920 [-] on Build GRCh38
          GenBankmRNA DA365821.1 - 1 245 Chr. 12 - 121796210 - 121802943 [-] on Build GRCh38
          GenBankmRNA DA505108.1 - 247 Chr. - - [-] on Build
          GenBankmRNA DT219909.1 - 1 247 Chr. 12 - 121796095 - 121802945 [-] on Build GRCh38
          GenBankmRNA H11978.1 - 2 140 Chr. 12 - 121799853 - 121802932 [-] on Build GRCh38
          GenBankmRNA BG699859.1 - 2 155 Chr. 12 - 121795399 - 121802942 [-] on Build GRCh38
          GenBankmRNA BX419131.2 - 1 229 Chr. 12 - 121796669 - 121802928 [-] on Build GRCh38
          GenBankmRNA BX344455.2 - 2 142 Chr. 12 - 121795769 - 121802930 [-] on Build GRCh38
          GenBankmRNA DT218208.1 - 2 153 Chr. 12 - 121796041 - 121802945 [-] on Build GRCh38
          GenBankmRNA DB137461.1 - 2 175 Chr. 12 - 121799553 - 121802945 [-] on Build GRCh38
          GenBankmRNA BE797083.1 - 2 114 Chr. 12 - 121795879 - 121802906 [-] on Build GRCh38
          GenBankmRNA BX473002.1 - 1 191 Chr. 12 - 121799871 - 121802889 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0