Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAXIP1-AS2 SIRNA Assays ›
          Gene Symbol:
          PAXIP1-AS2
          Gene Name:
          PAXIP1 antisense RNA 2
          Gene Aliases:
          PAXIP1OS
          Chromosome Location:
          Chr.7: 154928517 - 154949490 on Build GRCh38
          UniGene ID:
          Hs.586358
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260757

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PAXIP1-AS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024476.1 2 280
          NR_024477.1 2 280
          GenBank mRNA AK056609.1 - 2 270
          BC033053.1 - 2 306
          BC050402.1 - 2 265
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX377778.2 - 2 280 Chr. 7 - 154928517 - 154944724 [+] on Build GRCh38
          GenBankmRNA DA990117.1 - 2 291 Chr. 7 - 154928506 - 154944233 [+] on Build GRCh38
          GenBankmRNA DB161451.1 - 2 277 Chr. 7 - 154928520 - 154944284 [+] on Build GRCh38
          GenBankmRNA DA428625.1 - 2 332 Chr. 7 - 154928466 - 154944224 [+] on Build GRCh38
          GenBankmRNA BM549692.1 - 2 285 Chr. 7 - 154928516 - 154944617 [+] on Build GRCh38
          GenBankmRNA BE748183.1 - 274 Chr. - - [-] on Build
          GenBankmRNA DA640619.1 - 2 293 Chr. 7 - 154928504 - 154944230 [+] on Build GRCh38
          GenBankmRNA DA611646.1 - 2 302 Chr. 7 - 154928496 - 154944264 [+] on Build GRCh38
          GenBankmRNA DA421500.1 - 2 313 Chr. 7 - 154928484 - 154944249 [+] on Build GRCh38
          GenBankmRNA BF849788.1 - 2 145 Chr. 7 - 154928663 - 154944368 [+] on Build GRCh38
          GenBankmRNA DB013789.1 - 2 294 Chr. 7 - 154928504 - 154944273 [+] on Build GRCh38
          GenBankmRNA DA996509.1 - 2 272 Chr. 7 - 154928528 - 154944264 [+] on Build GRCh38
          GenBankmRNA DN997682.1 - 2 287 Chr. 7 - 154928519 - 154944247 [+] on Build GRCh38
          GenBankmRNA CA389913.1 - 2 142 Chr. 7 - 154928655 - 154946448 [+] on Build GRCh38
          GenBankmRNA DA268497.1 - 2 318 Chr. 7 - 154928481 - 154944236 [+] on Build GRCh38
          GenBankmRNA DA224698.1 - 2 307 Chr. 7 - 154928490 - 154944328 [+] on Build GRCh38
          GenBankmRNA DB217206.1 - 2 277 Chr. 7 - 154928520 - 154944203 [+] on Build GRCh38
          GenBankmRNA DA595771.1 - 2 278 Chr. 7 - 154928520 - 154944275 [+] on Build GRCh38
          GenBankmRNA DA761780.1 - 2 270 Chr. 7 - 154928528 - 154944283 [+] on Build GRCh38
          GenBankmRNA BX356348.2 - 2 280 Chr. 7 - 154928517 - 154944683 [+] on Build GRCh38
          GenBankmRNA DA252464.1 - 2 329 Chr. 7 - 154928469 - 154944252 [+] on Build GRCh38
          GenBankmRNA DB112724.1 - 2 272 Chr. 7 - 154928528 - 154944293 [+] on Build GRCh38
          GenBankmRNA BI545751.1 - 2 306 Chr. 7 - 154928494 - 154929006 [+] on Build GRCh38
          GenBankmRNA D81902.1 - 2 104 Chr. 7 - 154928693 - 154944371 [+] on Build GRCh38
          GenBankmRNA BF212893.1 - 2 347 Chr. 7 - 154928450 - 154930657 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0