Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100133050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027503.1 | 4 | 485 | |||
GenBank mRNA | BI830432.1 | - | 4 | 484 | ||
BQ109458.1 | - | 4 | 485 |
Gene Symbol: CTD-2151A2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110562.1 | 1 | 321 | |||
GenBank mRNA | CR742340.1 | - | 1 | 321 |