Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLK2P1 SIRNA Assays ›
          Gene Symbol:
          CLK2P1
          Gene Name:
          CDC like kinase 2, pseudogene 1
          Gene Aliases:
          CLK2P
          Chromosome Location:
          Chr.7: 23584716 - 23586527 on Build GRCh38
          UniGene ID:
          Hs.632635
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n259914

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLK2P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002711.3 1 251
          GenBank mRNA BC022025.1 - 1 267
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB054291.1 - 1 270 Chr. 7 - 23586052 - 23586546 [-] on Build GRCh38
          GenBankmRNA DB036023.1 - 1 238 Chr. 7 - 23585949 - 23586514 [-] on Build GRCh38
          GenBankmRNA DB038453.1 - 1 259 Chr. 7 - 23585969 - 23586534 [-] on Build GRCh38
          GenBankmRNA DB057698.1 - 1 258 Chr. 7 - 23586002 - 23586534 [-] on Build GRCh38
          GenBankmRNA DB471104.1 - 1 248 Chr. 7 - 23586050 - 23586524 [-] on Build GRCh38
          GenBankmRNA HY007936.1 - 1 251 Chr. 7 - 23585933 - 23586527 [-] on Build GRCh38
          GenBankmRNA DB446583.1 - 1 248 Chr. 7 - 23586097 - 23586524 [-] on Build GRCh38
          GenBankmRNA DB460497.1 - 1 262 Chr. 7 - 23586051 - 23586538 [-] on Build GRCh38
          GenBankmRNA HY049493.1 - 1 263 Chr. 7 - 23586111 - 23586538 [-] on Build GRCh38
          GenBankmRNA DB032385.1 - 1 276 Chr. 7 - 23585968 - 23586552 [-] on Build GRCh38
          GenBankmRNA DB023440.1 - 1 294 Chr. 7 - 23586020 - 23586570 [-] on Build GRCh38
          GenBankmRNA HY031637.1 - 1 277 Chr. 7 - 23586094 - 23586551 [-] on Build GRCh38
          GenBankmRNA DB022984.1 - 1 263 Chr. 7 - 23586052 - 23586538 [-] on Build GRCh38
          GenBankmRNA BM563567.1 - 240 Chr. - - [-] on Build
          GenBankmRNA DB443950.1 - 1 262 Chr. 7 - 23586087 - 23586538 [-] on Build GRCh38
          GenBankmRNA DB056472.1 - 1 262 Chr. 7 - 23586052 - 23586538 [-] on Build GRCh38
          GenBankmRNA HY021239.1 - 1 245 Chr. 7 - 23586048 - 23586520 [-] on Build GRCh38
          GenBankmRNA BG723070.1 - 1 237 Chr. 7 - 23585964 - 23586511 [-] on Build GRCh38
          GenBankmRNA BG717579.1 - 267 Chr. - - [-] on Build
          GenBankmRNA BG720946.1 - 1 266 Chr. 7 - 23585791 - 23586538 [-] on Build GRCh38
          GenBankmRNA DB056280.1 - 1 262 Chr. 7 - 23586052 - 23586538 [-] on Build GRCh38
          GenBankmRNA HY032935.1 - 1 234 Chr. 7 - 23586045 - 23586509 [-] on Build GRCh38
          GenBankmRNA HY015400.1 - 1 152 Chr. 7 - 23585892 - 23586428 [-] on Build GRCh38
          GenBankmRNA DB043867.1 - 1 263 Chr. 7 - 23585963 - 23586538 [-] on Build GRCh38
          GenBankmRNA HY031139.1 - 1 245 Chr. 7 - 23586058 - 23586520 [-] on Build GRCh38
          GenBankmRNA HY005607.1 - 1 245 Chr. 7 - 23586020 - 23586520 [-] on Build GRCh38
          GenBankmRNA DB458507.1 - 1 262 Chr. 7 - 23586061 - 23586538 [-] on Build GRCh38
          GenBankmRNA HY220942.1 - 1 199 Chr. 7 - 23586011 - 23586442 [-] on Build GRCh38
          GenBankmRNA DB032346.1 - 1 255 Chr. 7 - 23586026 - 23586531 [-] on Build GRCh38
          GenBankmRNA DB025100.1 - 1 262 Chr. 7 - 23585991 - 23586538 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0