Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF674-AS1 SIRNA Assays ›
          Gene Symbol:
          ZNF674-AS1
          Gene Name:
          ZNF674 antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 46545490 - 46548475 on Build GRCh38
          UniGene ID:
          Hs.729370
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258438

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF674-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015378.3 2 419
          GenBank mRNA AK056314.1 - 2 390
          BC034616.1 - 2 416
          DA707714.1 - 2 421
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AL554864.3 - 2 387 Chr. X - 46545522 - 46547393 [+] on Build GRCh38
          GenBankmRNA DA349434.1 - 2 414 Chr. X - 46545495 - 46546892 [+] on Build GRCh38
          GenBankmRNA DB008343.1 - 2 414 Chr. X - 46545496 - 46546844 [+] on Build GRCh38
          GenBankmRNA BX477590.1 - 2 393 Chr. X - 46545517 - 46546905 [+] on Build GRCh38
          GenBankmRNA BX488315.1 - 2 387 Chr. X - 46545523 - 46546906 [+] on Build GRCh38
          GenBankmRNA DA751230.1 - 2 390 Chr. X - 46545519 - 46546922 [+] on Build GRCh38
          GenBankmRNA DB124457.1 - 2 409 Chr. X - 46545500 - 46546887 [+] on Build GRCh38
          GenBankmRNA DA704174.1 - 2 390 Chr. X - 46545519 - 46546844 [+] on Build GRCh38
          GenBankmRNA BG719079.1 - 2 421 Chr. X - 46545493 - 46547106 [+] on Build GRCh38
          GenBankmRNA DA690322.1 - 2 409 Chr. X - 46545500 - 46546920 [+] on Build GRCh38
          GenBankmRNA CN419957.1 - 2 375 Chr. X - 46545534 - 46546935 [+] on Build GRCh38
          GenBankmRNA CD642058.1 - 2 400 Chr. X - 46545509 - 46547057 [+] on Build GRCh38
          GenBankmRNA DA355854.1 - 2 401 Chr. X - 46545508 - 46546898 [+] on Build GRCh38
          GenBankmRNA CN419958.1 - 2 375 Chr. X - 46545534 - 46547102 [+] on Build GRCh38
          GenBankmRNA DB267799.1 - 2 397 Chr. X - 46545512 - 46546917 [+] on Build GRCh38
          GenBankmRNA DA383874.1 - 2 390 Chr. X - 46545519 - 46546915 [+] on Build GRCh38
          GenBankmRNA DA750669.1 - 2 401 Chr. X - 46545508 - 46546917 [+] on Build GRCh38
          GenBankmRNA CK002253.1 - 2 401 Chr. X - 46545509 - 46546904 [+] on Build GRCh38
          GenBankmRNA BM547124.1 - 2 376 Chr. X - 46545552 - 46547418 [+] on Build GRCh38
          GenBankmRNA DA072641.1 - 2 397 Chr. X - 46545512 - 46546924 [+] on Build GRCh38
          GenBankmRNA DA377212.1 - 2 345 Chr. X - 46545564 - 46546967 [+] on Build GRCh38
          GenBankmRNA DB014058.1 - 2 403 Chr. X - 46545506 - 46546911 [+] on Build GRCh38
          GenBankmRNA DA026616.1 - 2 540 Chr. X - 46545370 - 46546800 [+] on Build GRCh38
          GenBankmRNA BX505213.1 - 2 382 Chr. X - 46545527 - 46546965 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0