Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 1700031M16RIK SIRNA Assays ›
          Gene Symbol:
          1700031M16Rik
          Gene Name:
          RIKEN cDNA 1700031M16 gene
          Gene Aliases:
          4930413L20Rik
          Chromosome Location:
          Chr.15: 98398445 - 98416135 on Build GRCm38
          UniGene ID:
          Mm.272760
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n256787

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 1700031M16Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015496.1 5 214
          GenBank mRNA AK006585.1 - 5 215
          AK015131.1 - 5 216
          BC048654.1 - 5 214
          BC048656.1 - 4 136
          BC099572.1 - 5 222
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF149638.1 - 4 160 Chr. 15 - 98399012 - 98415239 [+] on Build GRCm38
          GenBankmRNA CN833884.1 - 223 Chr. - - [-] on Build
          GenBankmRNA CA465480.1 - 5 217 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA BU962172.1 - 130 Chr. - - [-] on Build
          GenBankmRNA CN836046.1 - 5 217 Chr. 15 - 98398445 - 98415367 [+] on Build GRCm38
          GenBankmRNA BU935627.1 - 5 225 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA BU935618.1 - 5 224 Chr. 15 - 98398438 - 98415387 [+] on Build GRCm38
          GenBankmRNA CA465561.1 - 5 243 Chr. 15 - 98398419 - 98413590 [+] on Build GRCm38
          GenBankmRNA CN842460.1 - 225 Chr. - - [-] on Build
          GenBankmRNA CA463913.1 - 5 217 Chr. 15 - 98398445 - 98415387 [+] on Build GRCm38
          GenBankmRNA CA466332.1 - 5 217 Chr. 15 - 98398445 - 98415375 [+] on Build GRCm38
          GenBankmRNA BU962199.1 - 5 224 Chr. 15 - 98398438 - 98415388 [+] on Build GRCm38
          GenBankmRNA BU936479.1 - 217 Chr. - - [-] on Build
          GenBankmRNA BU961804.1 - 5 217 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA BU961612.1 - 4 130 Chr. 15 - 98398445 - 98415420 [+] on Build GRCm38
          GenBankmRNA BU936361.1 - 5 217 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA CA464881.1 - 5 216 Chr. 15 - 98398445 - 98415387 [+] on Build GRCm38
          GenBankmRNA BU935937.1 - 5 217 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA CA494517.1 - 4 139 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA BY706588.1 - 5 215 Chr. 15 - 98398445 - 98416273 [+] on Build GRCm38
          GenBankmRNA BU939195.1 - 216 Chr. - - [-] on Build
          GenBankmRNA CA463299.1 - 5 236 Chr. 15 - 98398424 - 98415389 [+] on Build GRCm38
          GenBankmRNA BU961616.1 - 217 Chr. - - [-] on Build
          GenBankmRNA CA494506.1 - 5 217 Chr. 15 - 98398445 - 98416116 [+] on Build GRCm38
          GenBankmRNA BU961355.1 - 5 217 Chr. 15 - 98398445 - 98415384 [+] on Build GRCm38
          GenBankmRNA CN832839.1 - 5 217 Chr. 15 - 98398445 - 98415344 [+] on Build GRCm38
          GenBankmRNA BU961403.1 - 5 217 Chr. 15 - 98398445 - 98415388 [+] on Build GRCm38
          GenBankmRNA BU961464.1 - 216 Chr. - - [-] on Build
          GenBankmRNA BY714676.1 - 5 216 Chr. 15 - 98398445 - 98412803 [+] on Build GRCm38
          GenBankmRNA BU936113.1 - 217 Chr. - - [-] on Build

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NR_036618, CK597145, CK652853.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0