Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEAT1 SIRNA Assays ›
          Gene Symbol:
          Neat1
          Gene Name:
          nuclear paraspeckle assembly transcript 1 (non-protein coding)
          Gene Aliases:
          2310043N10Rik, VINC
          Chromosome Location:
          Chr.19: 5842302 - 5845478 on Build GRCm38
          UniGene ID:
          Mm.281895
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n256282

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Neat1
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK018202.1 -
          1 938
          AK028745.1 - LIT3353
          VINC
          1 2574
          AK033579.1 -
          1 2572
          AK145725.1 -
          1 2572
          AK159400.1 -
          1 2571
          AK159773.1 -
          1 2574
          AK167154.1 -
          1 2573
          AK167255.1 -
          1 2572
          BC025215.1 -
          2 1927
          BC064019.1 -
          1 1795
          BC095948.1 -
          1 1752
          EF177378.1 -
          1 2571
          GQ859163.1 -
          2571
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CK625671.1 - 1 151 Chr. 19 - 5842469 - 5843058 [-] on Build GRCm38
          GenBankmRNA CB587823.1 - 1 113 Chr. 19 - 5842316 - 5843017 [-] on Build GRCm38
          GenBankmRNA BY757058.1 - 1 157 Chr. 19 - 5842302 - 5843055 [-] on Build GRCm38
          GenBankmRNA BB003096.2 - 1 32 Chr. 19 - 5842302 - 5842939 [-] on Build GRCm38
          GenBankmRNA AW913305.1 - 1 38 Chr. 19 - 5842413 - 5842945 [-] on Build GRCm38
          GenBankmRNA EH103182.1 - 1 38 Chr. 19 - 5842379 - 5842937 [-] on Build GRCm38
          GenBankmRNA BQ565654.1 - 1 114 Chr. 19 - 5842428 - 5843020 [-] on Build GRCm38
          GenBankmRNA BY756874.1 - 1 105 Chr. 19 - 5842302 - 5843012 [-] on Build GRCm38
          GenBankmRNA CJ227717.1 - 1 102 Chr. 19 - 5842309 - 5843009 [-] on Build GRCm38
          GenBankmRNA AA796610.1 - 1 100 Chr. 19 - 5842391 - 5842995 [-] on Build GRCm38
          GenBankmRNA BY759874.1 - 1 67 Chr. 19 - 5842309 - 5842974 [-] on Build GRCm38
          GenBankmRNA CK795780.1 - 1 220 Chr. 19 - 5842361 - 5843113 [-] on Build GRCm38
          GenBankmRNA CJ224670.1 - 1 110 Chr. 19 - 5842302 - 5843018 [-] on Build GRCm38
          GenBankmRNA BQ568729.1 - 1 113 Chr. 19 - 5842478 - 5843020 [-] on Build GRCm38
          GenBankmRNA CB575092.1 - 1 263 Chr. 19 - 5842305 - 5843170 [-] on Build GRCm38
          GenBankmRNA BI180688.1 - 1 98 Chr. 19 - 5842367 - 5842996 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0