Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEAT1 SIRNA Assays ›
          Gene Symbol:
          Neat1
          Gene Name:
          nuclear paraspeckle assembly transcript 1 (non-protein coding)
          Gene Aliases:
          2310043N10Rik, VINC
          Chromosome Location:
          Chr.19: 5842302 - 5845478 on Build GRCm38
          UniGene ID:
          Mm.281895
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n256277

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Neat1
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK028745.1 - LIT3353
          VINC
          1 203
          AK033579.1 -
          1 201
          AK145725.1 -
          1 201
          AK148054.1 -
          1 201
          AK159400.1 -
          1 200
          AK159773.1 -
          1 203
          AK167154.1 -
          1 202
          AK167255.1 -
          1 201
          EF177378.1 -
          1 200
          GQ859163.1 -
          200
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BB853648.1 - 1 201 Chr. 19 - 5845009 - 5845478 [-] on Build GRCm38
          GenBankmRNA BE292258.1 - 1 101 Chr. 19 - 5844819 - 5845379 [-] on Build GRCm38
          GenBankmRNA BY227756.1 - 1 199 Chr. 19 - 5845076 - 5845477 [-] on Build GRCm38
          GenBankmRNA BG975159.1 - 1 187 Chr. 19 - 5844763 - 5845466 [-] on Build GRCm38
          GenBankmRNA BI662017.1 - 1 184 Chr. 19 - 5844568 - 5845463 [-] on Build GRCm38
          GenBankmRNA BY289413.1 - 1 201 Chr. 19 - 5845036 - 5845478 [-] on Build GRCm38
          GenBankmRNA CJ092781.1 - 1 201 Chr. 19 - 5844863 - 5845478 [-] on Build GRCm38
          GenBankmRNA CF905600.1 - 1 89 Chr. 19 - 5844836 - 5845367 [-] on Build GRCm38
          GenBankmRNA CK791098.1 - 1 179 Chr. 19 - 5844671 - 5845457 [-] on Build GRCm38
          GenBankmRNA BI687086.1 - 1 185 Chr. 19 - 5844678 - 5845463 [-] on Build GRCm38
          GenBankmRNA BY287929.1 - 1 201 Chr. 19 - 5845072 - 5845478 [-] on Build GRCm38
          GenBankmRNA CA543689.1 - 1 185 Chr. 19 - 5845059 - 5845463 [-] on Build GRCm38
          GenBankmRNA CF164448.1 - 1 17 Chr. 19 - 5845058 - 5845295 [-] on Build GRCm38
          GenBankmRNA BB854946.1 - 1 200 Chr. 19 - 5844983 - 5845477 [-] on Build GRCm38
          GenBankmRNA BI684752.1 - 1 200 Chr. 19 - 5844826 - 5845468 [-] on Build GRCm38
          GenBankmRNA CX220447.1 - 1 180 Chr. 19 - 5844850 - 5845458 [-] on Build GRCm38
          GenBankmRNA CF905509.1 - 1 190 Chr. 19 - 5844832 - 5845468 [-] on Build GRCm38
          GenBankmRNA BF788715.1 - 1 175 Chr. 19 - 5844660 - 5845449 [-] on Build GRCm38
          GenBankmRNA BB858090.1 - 1 201 Chr. 19 - 5845003 - 5845478 [-] on Build GRCm38
          GenBankmRNA BB851653.1 - 1 201 Chr. 19 - 5844998 - 5845477 [-] on Build GRCm38
          GenBankmRNA CA491124.1 - 237 Chr. - - [-] on Build
          GenBankmRNA BI251714.1 - 1 151 Chr. 19 - 5844642 - 5845430 [-] on Build GRCm38
          GenBankmRNA AI747399.1 - 333 Chr. - - [-] on Build
          GenBankmRNA BB853323.1 - 1 201 Chr. 19 - 5844980 - 5845478 [-] on Build GRCm38
          GenBankmRNA BI663474.1 - 1 185 Chr. 19 - 5844435 - 5845441 [-] on Build GRCm38
          GenBankmRNA BF787265.1 - 1 213 Chr. 19 - 5844906 - 5845466 [-] on Build GRCm38
          GenBankmRNA BE570034.1 - 1 163 Chr. 19 - 5844861 - 5845442 [-] on Build GRCm38
          GenBankmRNA BY228532.1 - 1 201 Chr. 19 - 5845066 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY070849.1 - 1 201 Chr. 19 - 5845082 - 5845478 [-] on Build GRCm38
          GenBankmRNA BU696061.1 - 1 190 Chr. 19 - 5844893 - 5845459 [-] on Build GRCm38
          GenBankmRNA BB856558.1 - 1 201 Chr. 19 - 5845001 - 5845478 [-] on Build GRCm38
          GenBankmRNA BQ922797.1 - 1 175 Chr. 19 - 5844637 - 5845453 [-] on Build GRCm38
          GenBankmRNA CX206360.1 - 1 91 Chr. 19 - 5844771 - 5845369 [-] on Build GRCm38
          GenBankmRNA BY226478.1 - 1 201 Chr. 19 - 5845044 - 5845477 [-] on Build GRCm38
          GenBankmRNA CK788118.1 - 1 183 Chr. 19 - 5844698 - 5845461 [-] on Build GRCm38
          GenBankmRNA BB857209.1 - 1 201 Chr. 19 - 5844997 - 5845478 [-] on Build GRCm38
          GenBankmRNA BB855765.1 - 1 201 Chr. 19 - 5844982 - 5845478 [-] on Build GRCm38
          GenBankmRNA BB854544.1 - 1 201 Chr. 19 - 5845024 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY227940.1 - 1 201 Chr. 19 - 5845050 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY228305.1 - 1 201 Chr. 19 - 5845053 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY237486.1 - 1 201 Chr. 19 - 5845069 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY754186.1 - 1 203 Chr. 19 - 5844842 - 5845480 [-] on Build GRCm38
          GenBankmRNA AW320633.1 - 1 157 Chr. 19 - 5844970 - 5845435 [-] on Build GRCm38
          GenBankmRNA BY228830.1 - 1 203 Chr. 19 - 5845069 - 5845480 [-] on Build GRCm38
          GenBankmRNA BF785338.1 - 1 190 Chr. 19 - 5844682 - 5845468 [-] on Build GRCm38
          GenBankmRNA BY738983.1 - 1 201 Chr. 19 - 5844819 - 5845478 [-] on Build GRCm38
          GenBankmRNA CA544297.1 - 1 192 Chr. 19 - 5844985 - 5845470 [-] on Build GRCm38
          GenBankmRNA CF903281.1 - 1 118 Chr. 19 - 5844858 - 5845396 [-] on Build GRCm38
          GenBankmRNA CJ092647.1 - 1 203 Chr. 19 - 5844863 - 5845480 [-] on Build GRCm38
          GenBankmRNA CF904213.1 - 1 108 Chr. 19 - 5844761 - 5845386 [-] on Build GRCm38
          GenBankmRNA BF583956.1 - 193 Chr. - - [-] on Build
          GenBankmRNA CN667222.1 - 1 190 Chr. 19 - 5844954 - 5845468 [-] on Build GRCm38
          GenBankmRNA CF903604.1 - 1 104 Chr. 19 - 5844793 - 5845382 [-] on Build GRCm38
          GenBankmRNA BB856230.1 - 1 200 Chr. 19 - 5845017 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY736686.1 - 1 201 Chr. 19 - 5844864 - 5845478 [-] on Build GRCm38
          GenBankmRNA BF117997.1 - 1 167 Chr. 19 - 5844987 - 5845436 [-] on Build GRCm38
          GenBankmRNA BI455008.1 - 1 190 Chr. 19 - 5844717 - 5845468 [-] on Build GRCm38
          GenBankmRNA BG918357.1 - 1 181 Chr. 19 - 5844762 - 5845459 [-] on Build GRCm38
          GenBankmRNA BI248695.1 - 1 198 Chr. 19 - 5844706 - 5845467 [-] on Build GRCm38
          GenBankmRNA BG917546.1 - 1 144 Chr. 19 - 5844789 - 5845422 [-] on Build GRCm38
          GenBankmRNA BB857218.1 - 1 88 Chr. 19 - 5844891 - 5845366 [-] on Build GRCm38
          GenBankmRNA CJ089083.1 - 1 202 Chr. 19 - 5844762 - 5845480 [-] on Build GRCm38
          GenBankmRNA BI662952.1 - 1 185 Chr. 19 - 5844829 - 5845463 [-] on Build GRCm38
          GenBankmRNA BI082923.1 - 1 66 Chr. 19 - 5844812 - 5845343 [-] on Build GRCm38
          GenBankmRNA BB613043.1 - 1 201 Chr. 19 - 5844839 - 5845478 [-] on Build GRCm38
          GenBankmRNA BB851976.1 - 1 201 Chr. 19 - 5845017 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY226738.1 - 1 201 Chr. 19 - 5845066 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY275505.1 - 1 201 Chr. 19 - 5845066 - 5845478 [-] on Build GRCm38
          GenBankmRNA AA839206.1 - 1 102 Chr. 19 - 5844944 - 5845380 [-] on Build GRCm38
          GenBankmRNA BB625113.1 - 1 201 Chr. 19 - 5844808 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY736500.1 - 1 201 Chr. 19 - 5844765 - 5845478 [-] on Build GRCm38
          GenBankmRNA AI786586.1 - 1 158 Chr. 19 - 5844918 - 5845436 [-] on Build GRCm38
          GenBankmRNA AI317792.1 - 1 199 Chr. 19 - 5844915 - 5845477 [-] on Build GRCm38
          GenBankmRNA BY227743.1 - 1 201 Chr. 19 - 5845079 - 5845478 [-] on Build GRCm38
          GenBankmRNA CA567078.1 - 1 129 Chr. 19 - 5844965 - 5845407 [-] on Build GRCm38
          GenBankmRNA BF686899.1 - 202 Chr. - - [-] on Build
          GenBankmRNA BB854684.1 - 1 201 Chr. 19 - 5845015 - 5845478 [-] on Build GRCm38
          GenBankmRNA CA566714.1 - 1 152 Chr. 19 - 5844967 - 5845430 [-] on Build GRCm38
          GenBankmRNA BI409246.1 - 1 149 Chr. 19 - 5844626 - 5845414 [-] on Build GRCm38
          GenBankmRNA BB855243.1 - 1 201 Chr. 19 - 5845009 - 5845478 [-] on Build GRCm38
          GenBankmRNA BI665912.1 - 1 183 Chr. 19 - 5844701 - 5845460 [-] on Build GRCm38
          GenBankmRNA BY753750.1 - 1 200 Chr. 19 - 5844857 - 5845476 [-] on Build GRCm38
          GenBankmRNA BG976423.1 - 1 187 Chr. 19 - 5844740 - 5845464 [-] on Build GRCm38
          GenBankmRNA BQ267061.1 - 1 197 Chr. 19 - 5844933 - 5845463 [-] on Build GRCm38
          GenBankmRNA BF578400.1 - 162 Chr. - - [-] on Build
          GenBankmRNA BG918993.1 - 1 185 Chr. 19 - 5844783 - 5845463 [-] on Build GRCm38
          GenBankmRNA BI689272.1 - 1 168 Chr. 19 - 5844616 - 5845446 [-] on Build GRCm38
          GenBankmRNA BB852115.1 - 1 201 Chr. 19 - 5845018 - 5845478 [-] on Build GRCm38
          GenBankmRNA BY288284.1 - 1 201 Chr. 19 - 5845068 - 5845478 [-] on Build GRCm38
          GenBankmRNA AL364248.1 - 1 45 Chr. 19 - 5844926 - 5845323 [-] on Build GRCm38
          GenBankmRNA BY289412.1 - 1 201 Chr. 19 - 5845036 - 5845478 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0