Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 4933439C10RIK SIRNA Assays ›
          Gene Symbol:
          4933439C10Rik
          Gene Name:
          RIKEN cDNA 4933439C10 gene
          Gene Aliases:
          2610028M21Rik
          Chromosome Location:
          Chr.11: 59505685 - 59511067 on Build GRCm38
          UniGene ID:
          Mm.440257
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n254034

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 4933439C10Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015585.2 4 1481
          GenBank mRNA AK011602.1 - 4 1014
          AK017115.1 - 4 1482
          AK030920.1 - 1509
          AK051100.1 - 2 2732
          AK163429.1 - 4 1477
          AK169756.1 - 4 1511
          BC065402.1 - 3 187
          BC099591.1 - 4 728
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ928558.1 - 4 487 Chr. 11 - 59506679 - 59510106 [+] on Build GRCm38
          GenBankmRNA BX525008.1 - 4 394 Chr. 11 - 59506791 - 59510846 [+] on Build GRCm38
          GenBankmRNA BY762673.1 - 3 296 Chr. 11 - 59507190 - 59511067 [+] on Build GRCm38
          GenBankmRNA AI036408.1 - 3 173 Chr. 11 - 59507641 - 59511012 [+] on Build GRCm38
          GenBankmRNA CJ317149.1 - 1 49 Chr. 11 - 59509654 - 59511070 [+] on Build GRCm38
          GenBankmRNA EH098200.1 - 2 155 Chr. 11 - 59507842 - 59511069 [+] on Build GRCm38
          GenBankmRNA BY754733.1 - 3 360 Chr. 11 - 59507225 - 59510103 [+] on Build GRCm38
          GenBankmRNA W98666.1 - 377 Chr. - - [-] on Build
          GenBankmRNA EH099787.1 - 3 296 Chr. 11 - 59507189 - 59510930 [+] on Build GRCm38
          GenBankmRNA CX243337.1 - 4 403 Chr. 11 - 59506763 - 59510086 [+] on Build GRCm38
          GenBankmRNA BB802259.1 - 1 14 Chr. 11 - 59509689 - 59511067 [+] on Build GRCm38
          GenBankmRNA BE133382.1 - 3 187 Chr. 11 - 59507283 - 59510940 [+] on Build GRCm38
          GenBankmRNA EH102917.1 - 3 296 Chr. 11 - 59507189 - 59511035 [+] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-854c5b95bd-957wg:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0